Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010840.hg.1 |
ENST00000589259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010840.hg.1 |
ENST00000589259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010840.hg.1 |
ENST00000589259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010840.hg.1 |
ENST00000589259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010840.hg.1 |
ENST00000589259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010840.hg.1 |
NR_110661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010840.hg.1 |
NR_110661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010840.hg.1 |
NR_110661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010840.hg.1 |
NR_110661.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010840.hg.1 |
NR_110661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700186788.hg.1 |
ENST00000589259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700186788.hg.1 |
NR_110661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700186788.hg.1 |
TC1700010840.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700186788.hg.1 |
NR_110661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700186788.hg.1 |
ENST00000589259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700186787.hg.1 |
ENST00000589259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700186787.hg.1 |
NR_110661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700186787.hg.1 |
TC1700010840.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700186787.hg.1 |
ENST00000589259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700186787.hg.1 |
NR_110661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010840.hg.1 |
ENST00000589259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010840.hg.1 |
ENST00000589259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010840.hg.1 |
ENST00000589259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010840.hg.1 |
ENST00000589259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010840.hg.1 |
ENST00000589259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010840.hg.1 |
NR_110661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010840.hg.1 |
NR_110661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010840.hg.1 |
NR_110661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010840.hg.1 |
NR_110661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010840.hg.1 |
NR_110661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700186790.hg.1 |
ENST00000589259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700186790.hg.1 |
NR_110661.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700186790.hg.1 |
TC1700010840.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700186790.hg.1 |
ENST00000589259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700186790.hg.1 |
NR_110661.1 |
Refseq |