Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011616.hg.1 |
ENST00000456026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011616.hg.1 |
ENST00000456026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011616.hg.1 |
ENST00000456026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011616.hg.1 |
ENST00000456026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011616.hg.1 |
ENST00000456026.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011616.hg.1 |
NR_110794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011616.hg.1 |
NR_110794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011616.hg.1 |
NR_110794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011616.hg.1 |
NR_110794.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011616.hg.1 |
NR_110794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185523.hg.1 |
ENST00000456026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185523.hg.1 |
NR_110794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185523.hg.1 |
TC0100011616.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100185523.hg.1 |
NR_110794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185523.hg.1 |
ENST00000456026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185525.hg.1 |
ENST00000456026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185525.hg.1 |
NR_110794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185525.hg.1 |
TC0100011616.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100185525.hg.1 |
ENST00000456026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185525.hg.1 |
NR_110794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185526.hg.1 |
ENST00000456026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185526.hg.1 |
NR_110794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185526.hg.1 |
TC0100011616.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100185526.hg.1 |
ENST00000456026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185526.hg.1 |
NR_110794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011616.hg.1 |
ENST00000456026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011616.hg.1 |
ENST00000456026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011616.hg.1 |
ENST00000456026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011616.hg.1 |
ENST00000456026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011616.hg.1 |
ENST00000456026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011616.hg.1 |
NR_110794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011616.hg.1 |
NR_110794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011616.hg.1 |
NR_110794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011616.hg.1 |
NR_110794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011616.hg.1 |
NR_110794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185524.hg.1 |
ENST00000456026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185524.hg.1 |
NR_110794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185524.hg.1 |
TC0100011616.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100185524.hg.1 |
ENST00000456026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185524.hg.1 |
NR_110794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185522.hg.1 |
ENST00000456026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185522.hg.1 |
NR_110794.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100185522.hg.1 |
TC0100011616.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100185522.hg.1 |
ENST00000456026.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100185522.hg.1 |
NR_110794.1 |
Refseq |