Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008876.hg.1 |
ENST00000577295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008876.hg.1 |
ENST00000577295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008876.hg.1 |
ENST00000577295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008876.hg.1 |
ENST00000577295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008876.hg.1 |
ENST00000577295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008876.hg.1 |
NR_110878.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008876.hg.1 |
NR_110878.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008876.hg.1 |
NR_110878.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008876.hg.1 |
NR_110878.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008876.hg.1 |
NR_110878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166327.hg.1 |
ENST00000577295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166327.hg.1 |
NR_110878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166327.hg.1 |
TC1700008876.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700166327.hg.1 |
NR_110878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166327.hg.1 |
ENST00000577295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166324.hg.1 |
ENST00000577295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166324.hg.1 |
NR_110878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166324.hg.1 |
TC1700008876.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700166324.hg.1 |
ENST00000577295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166324.hg.1 |
NR_110878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166328.hg.1 |
ENST00000577295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166328.hg.1 |
NR_110878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166328.hg.1 |
TC1700008876.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700166328.hg.1 |
ENST00000577295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166328.hg.1 |
NR_110878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166325.hg.1 |
ENST00000577295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166325.hg.1 |
NR_110878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166325.hg.1 |
TC1700008876.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700166325.hg.1 |
ENST00000577295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166325.hg.1 |
NR_110878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008876.hg.1 |
ENST00000577295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008876.hg.1 |
ENST00000577295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008876.hg.1 |
ENST00000577295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008876.hg.1 |
ENST00000577295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008876.hg.1 |
ENST00000577295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008876.hg.1 |
NR_110878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008876.hg.1 |
NR_110878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008876.hg.1 |
NR_110878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008876.hg.1 |
NR_110878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008876.hg.1 |
NR_110878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166326.hg.1 |
ENST00000577295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166326.hg.1 |
NR_110878.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700166326.hg.1 |
TC1700008876.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700166326.hg.1 |
ENST00000577295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700166326.hg.1 |
NR_110878.1 |
Refseq |