Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009160.hg.1 |
ENST00000513093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009160.hg.1 |
ENST00000513093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009160.hg.1 |
ENST00000513093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009160.hg.1 |
ENST00000513093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009160.hg.1 |
ENST00000513093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009160.hg.1 |
ENST00000513093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009160.hg.1 |
NR_125888.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009160.hg.1 |
NR_125888.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009160.hg.1 |
NR_125888.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009160.hg.1 |
NR_125888.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009160.hg.1 |
NR_125888.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009160.hg.1 |
NR_125888.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163086.hg.1 |
ENST00000513093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163086.hg.1 |
NR_125888.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163086.hg.1 |
TC0400009160.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400163086.hg.1 |
NR_125888.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163086.hg.1 |
ENST00000513093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009160.hg.1 |
ENST00000513093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009160.hg.1 |
ENST00000513093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009160.hg.1 |
ENST00000513093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009160.hg.1 |
ENST00000513093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009160.hg.1 |
ENST00000513093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009160.hg.1 |
ENST00000513093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009160.hg.1 |
NR_125888.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009160.hg.1 |
NR_125888.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009160.hg.1 |
NR_125888.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009160.hg.1 |
NR_125888.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009160.hg.1 |
NR_125888.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009160.hg.1 |
NR_125888.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163088.hg.1 |
ENST00000513093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163088.hg.1 |
NR_125888.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163088.hg.1 |
TC0400009160.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400163088.hg.1 |
ENST00000513093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163088.hg.1 |
NR_125888.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163090.hg.1 |
ENST00000513093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163090.hg.1 |
NR_125888.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163090.hg.1 |
TC0400009160.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400163090.hg.1 |
ENST00000513093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163090.hg.1 |
NR_125888.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163085.hg.1 |
ENST00000513093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163085.hg.1 |
NR_125888.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163085.hg.1 |
TC0400009160.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400163085.hg.1 |
ENST00000513093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163085.hg.1 |
NR_125888.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163087.hg.1 |
ENST00000513093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163087.hg.1 |
NR_125888.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163087.hg.1 |
TC0400009160.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400163087.hg.1 |
ENST00000513093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163087.hg.1 |
NR_125888.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163089.hg.1 |
ENST00000513093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163089.hg.1 |
NR_125888.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163089.hg.1 |
TC0400009160.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400163089.hg.1 |
ENST00000513093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163089.hg.1 |
NR_125888.1 |
Refseq |