Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007216.hg.1 |
ENST00000603514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007216.hg.1 |
ENST00000603514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007216.hg.1 |
ENST00000603514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007216.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007216.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007216.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007216.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007216.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007216.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007216.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007216.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007216.hg.1 |
NR_104632.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007216.hg.1 |
NR_104632.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007216.hg.1 |
NR_104632.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007216.hg.1 |
NR_104632.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148792.hg.1 |
ENST00000603514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148792.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148792.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148792.hg.1 |
NR_104632.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148792.hg.1 |
TC0500007216.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500148792.hg.1 |
NR_104632.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148792.hg.1 |
ENST00000603514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148792.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148792.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148793.hg.1 |
ENST00000603514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148793.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148793.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148793.hg.1 |
NR_104632.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148793.hg.1 |
TC0500007216.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500148793.hg.1 |
ENST00000603514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148793.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148793.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148793.hg.1 |
NR_104632.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148788.hg.1 |
ENST00000603514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148788.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148788.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148788.hg.1 |
NR_104632.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148788.hg.1 |
TC0500007216.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500148788.hg.1 |
ENST00000603514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148788.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148788.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148788.hg.1 |
NR_104632.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007216.hg.1 |
ENST00000603514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007216.hg.1 |
ENST00000603514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007216.hg.1 |
ENST00000603514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007216.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007216.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007216.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007216.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007216.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007216.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007216.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007216.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007216.hg.1 |
NR_104632.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007216.hg.1 |
NR_104632.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007216.hg.1 |
NR_104632.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007216.hg.1 |
NR_104632.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148786.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148786.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148786.hg.1 |
NR_104632.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148786.hg.1 |
TC0500007216.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500148786.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148786.hg.1 |
ENST00000604954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148786.hg.1 |
NR_104632.1 |
Refseq |