Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
ENST00000551021.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
ENST00000551021.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
ENST00000551021.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
ENST00000551021.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
ENST00000551021.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
ENST00000551021.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
ENST00000551503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
ENST00000551503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
ENST00000551503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
ENST00000551503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
ENST00000551503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
ENST00000551503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
ENST00000551503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
ENST00000551503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
ENST00000551503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
ENST00000551503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
ENST00000607353.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
ENST00000607353.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
ENST00000607353.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
ENST00000607353.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
ENST00000607353.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
NR_125377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
NR_125377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
NR_125377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
NR_125377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
NR_125377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
NR_125377.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
NR_125378.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
NR_125378.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
NR_125378.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
NR_125378.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
NR_125378.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
NR_125379.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
NR_125379.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
NR_125379.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
NR_125379.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
NR_125380.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
NR_125380.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
NR_125380.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
NR_125380.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
NR_125381.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
NR_125381.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
NR_125381.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012618.hg.1 |
NR_125381.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200199724.hg.1 |
ENST00000551503.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200199724.hg.1 |
ENST00000551503.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200199724.hg.1 |
NR_125378.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200199724.hg.1 |
TC1200012618.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200199724.hg.1 |
NR_125378.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200199724.hg.1 |
ENST00000551503.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200199724.hg.1 |
ENST00000551503.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
ENST00000551021.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
ENST00000551021.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
ENST00000551021.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
ENST00000551021.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
ENST00000551021.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
ENST00000551021.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
ENST00000551503.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
ENST00000551503.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
ENST00000551503.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
ENST00000551503.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
ENST00000551503.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
ENST00000551503.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
ENST00000551503.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
ENST00000551503.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
ENST00000551503.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
ENST00000551503.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
ENST00000607353.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
ENST00000607353.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
ENST00000607353.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
ENST00000607353.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
ENST00000607353.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
NR_125377.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
NR_125377.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
NR_125377.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
NR_125377.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
NR_125377.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
NR_125377.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
NR_125378.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
NR_125378.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
NR_125378.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
NR_125378.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
NR_125378.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
NR_125379.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
NR_125379.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
NR_125379.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
NR_125379.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
NR_125380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
NR_125380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
NR_125380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
NR_125380.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
NR_125381.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
NR_125381.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
NR_125381.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012618.hg.1 |
NR_125381.1 |
Refseq |