Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04912738_m1
          See other SMG7 GE Assays ›
          Gene Symbol:
          SMG7
          Gene Name:
          SMG7, nonsense mediated mRNA decay factor
          Gene Aliases:
          C1orf16, EST1C, SGA56M
          Chromosome Location:
          Chr.1: 183471962 - 183554193 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04912738_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SMG7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173156.2 NP_775179.1 2 - 3 313 74
          NM_201568.2 NP_963862.1 2 - 3 313 74
          NM_201569.2 NP_963863.2 2 - 3 313 74
          XM_005245648.3 XP_005245705.1 4 - 5 998 74
          XM_005245649.3 XP_005245706.1 3 - 4 861 74
          XM_005245652.3 XP_005245709.1 2 - 3 182 74
          XM_005245653.4 XP_005245710.1 3 - 4 284 74
          XM_006711676.3 XP_006711739.1 3 - 4 861 74
          XM_006711678.3 XP_006711741.1 2 - 3 182 74
          XM_011510205.2 XP_011508507.1 5 - 6 1176 74
          XM_011510206.2 XP_011508508.1 5 - 6 1176 74
          XM_011510207.2 XP_011508509.1 3 - 4 900 74
          XM_011510208.2 XP_011508510.1 5 - 6 1176 74
          XM_011510209.2 XP_011508511.1 5 - 6 1176 74
          XM_017002966.1 XP_016858455.1 4 - 5 453 74
          XM_017002967.1 XP_016858456.1 4 - 5 461 74
          XM_017002969.1 XP_016858458.1 2 - 3 313 74
          XM_017002970.1 XP_016858459.1 2 - 3 313 74
          XM_017002971.1 XP_016858460.1 2 - 3 313 74
          GenBank mRNA AB085674.1 - 2 - 3 126 74
          AK056035.1 - 2 - 3 182 74
          AK074297.1 - 2 - 3 170 74
          AK297207.1 - 3 - 4 345 74
          AK307394.1 - 2 - 3 174 74
          BC036381.1 - 2 - 3 181 4829267 74
          BM152501.1 - 2 - 3 186 74
          D87437.2 - 2 - 3 157 74
          DB223038.1 - 2 - 3 313 74
          DC380923.1 - 2 - 3 295 74

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the XM_005245653.4, XM_011510207.2, XM_017002969.1, XM_017002970.1, XM_017002971.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SMG7

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
          mRNA export from nucleus
          regulation of dephosphorylation
          protein binding
          telomeric DNA binding
          protein phosphatase 2A binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0