Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00972 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_134240.1 | 3 - 4 | 381 | 97 | ||
GenBank mRNA | BG220912.1 | - | 3 - 4 | 381 | 97 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | semaphorin 3D | SEMA3D |
Sema-Z2, coll-2 |
-288.1kb | Anti-sense | Chr. 7 - 84995192 - 85187176 on NCBI Build GRCh38 |
Downstream | glutamate metabotropic receptor 3 | GRM3 |
GLUR3, GPRC1C, MGLUR3, mGlu3 |
1.2mb | Sense | Chr. 7 - 86643914 - 86864879 on NCBI Build GRCh38 |
Set Membership: |
![]() ![]() ![]() ![]() |