Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04938122_m1
          See other BCL2L1 GE Assays ›
          Gene Symbol:
          BCL2L1
          Gene Name:
          BCL2 like 1
          Gene Aliases:
          BCL-XL/S, BCL2L, BCLX, Bcl-X, PPP1R52
          Chromosome Location:
          Chr.20: 31664452 - 31723999 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04938122_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BCL2L1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_134257.1 2 - 3 831 72
          GenBank mRNA BX647525.1 - 4 - 5 709 72
          CD359906.1 - 2 - 3 447 72
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BQ691032.1 - 1 - 2 168 Chr. 20 - 31665393 - 31667148 [-] on Build GRCh38 72
          GenBankmRNA BM043798.1 - 1 - 2 46 Chr. 20 - 31665568 - 31667037 [-] on Build GRCh38 72
          GenBankmRNA BE393580.1 - 2 - 3 288 Chr. 20 - 31665822 - 31721787 [-] on Build GRCh38 72
          GenBankmRNA BQ943707.1 - 1 - 2 44 Chr. 20 - 31665402 - 31667036 [-] on Build GRCh38 72

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream cytochrome c oxidase subunit 4I2 COX4I2 COX4,
          COX4-2,
          COX4B,
          COX4L2,
          COXIV-2,
          dJ857M17.2
          -19.4kb Anti-sense Chr. 20 - 31637856 - 31645009 on NCBI Build GRCh38
          Downstream apoptotic BCL2L1-antisense long non-coding RNA ABALON INXS 54.4kb Anti-sense Chr. 20 - 31721507 - 31723409 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          cytokinesis
          ovarian follicle development
          in utero embryonic development
          release of cytochrome c from mitochondria
          endocytosis
          mitotic cell cycle checkpoint signaling
          germ cell development
          spermatogenesis
          cell population proliferation
          positive regulation of cell population proliferation
          male gonad development
          intrinsic apoptotic signaling pathway in response to DNA damage
          apoptotic mitochondrial changes
          fertilization
          suppression by virus of host apoptotic process
          response to cytokine
          growth
          negative regulation of apoptotic process
          negative regulation of neuron apoptotic process
          response to cycloheximide
          regulation of mitochondrial membrane permeability
          neuron apoptotic process
          regulation of mitochondrial membrane potential
          mitochondrion morphogenesis
          cellular response to amino acid stimulus
          cellular response to alkaloid
          cellular response to gamma radiation
          apoptotic process in bone marrow cell
          negative regulation of release of cytochrome c from mitochondria
          extrinsic apoptotic signaling pathway in absence of ligand
          hepatocyte apoptotic process
          negative regulation of execution phase of apoptosis
          negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
          negative regulation of protein localization to plasma membrane
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          negative regulation of intrinsic apoptotic signaling pathway
          positive regulation of intrinsic apoptotic signaling pathway
          protein binding
          protein kinase binding
          identical protein binding
          protein homodimerization activity
          protein heterodimerization activity
          BH3 domain binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix