Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105372069 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_134585.1 | 3 - 4 | 259 | 160 | ||
GenBank mRNA | BC037214.1 | - | 3 - 4 | 259 | 160 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BG721716.1 | - | 264 | Chr. - - [-] on Build | 160 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | KIAA1328 | KIAA1328 | Anti-sense | Chr. 18 - 36828929 - 37232127 on NCBI Build GRCh38 | ||
Downstream |
CUGBP, Elav-like family member 4 |
CELF4 |
BRUNOL4, CELF-4 |
339.4kb | Sense | Chr. 18 - 37243040 - 37566052 on NCBI Build GRCh38 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |