Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012825.hg.1 |
ENST00000597259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012825.hg.1 |
ENST00000597259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012825.hg.1 |
ENST00000598772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012825.hg.1 |
ENST00000598772.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400012825.hg.1 |
ENST00000598772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012825.hg.1 |
ENST00000597259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012825.hg.1 |
ENST00000597259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012825.hg.1 |
ENST00000598772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012825.hg.1 |
ENST00000598772.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400012825.hg.1 |
ENST00000598772.1 |
Ensembl |