Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04938552_m1
          See other LOC645553 GE Assays ›
          Gene Symbol:
          LOC645553
          Gene Name:
          uncharacterized LOC645553
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 46189029 - 46203062 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04938552_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC645553
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_135234.1 1 - 2 224 72
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DW418539.1 - 1 - 2 138 Chr. 19 - 46198157 - 46202958 [-] on Build GRCh38 72
          GenBankmRNA DW421781.1 - 1 - 2 262 Chr. 19 - 46198408 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW429675.1 - 1 - 2 262 Chr. 19 - 46198306 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW412438.1 - 1 - 2 261 Chr. 19 - 46198379 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW407847.1 - 1 - 2 262 Chr. 19 - 46198239 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW414954.1 - 1 - 2 261 Chr. 19 - 46198389 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW409751.1 - 1 - 2 261 Chr. 19 - 46198366 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW464774.1 - 1 - 2 262 Chr. 19 - 46198342 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW465474.1 - 1 - 2 262 Chr. 19 - 46198347 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW414911.1 - 1 - 2 262 Chr. 19 - 46198390 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW441009.1 - 1 - 2 262 Chr. 19 - 46198323 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW407868.1 - 1 - 2 262 Chr. 19 - 46198243 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW408304.1 - 1 - 2 262 Chr. 19 - 46198360 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW429975.1 - 1 - 2 262 Chr. 19 - 46198311 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW417340.1 - 1 - 2 262 Chr. 19 - 46198398 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW465626.1 - 1 - 2 262 Chr. 19 - 46198353 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW418712.1 - 1 - 2 262 Chr. 19 - 46198286 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW408791.1 - 1 - 2 262 Chr. 19 - 46198363 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW462466.1 - 1 - 2 262 Chr. 19 - 46198336 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW419170.1 - 1 - 2 262 Chr. 19 - 46198297 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW411996.1 - 2 - 3 124 Chr. 19 - 46198240 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW467586.1 - 1 - 2 262 Chr. 19 - 46198359 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW408303.1 - 1 - 2 262 Chr. 19 - 46198360 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW440420.1 - 1 - 2 262 Chr. 19 - 46198317 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW430121.1 - 1 - 2 262 Chr. 19 - 46198313 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW409935.1 - 1 - 2 118 Chr. 19 - 46198221 - 46202939 [-] on Build GRCh38 72
          GenBankmRNA DW414337.1 - 1 - 2 262 Chr. 19 - 46198388 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW412647.1 - 1 - 2 262 Chr. 19 - 46198381 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW412402.1 - 1 - 2 262 Chr. 19 - 46198380 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW418858.1 - 1 - 2 262 Chr. 19 - 46198290 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW418633.1 - 1 - 2 262 Chr. 19 - 46198284 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW416383.1 - 1 - 2 262 Chr. 19 - 46198403 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW413613.1 - 1 - 2 262 Chr. 19 - 46198385 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW451821.1 - 1 - 2 261 Chr. 19 - 46198329 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW418934.1 - 1 - 2 262 Chr. 19 - 46198292 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW412301.1 - 1 - 2 172 Chr. 19 - 46198289 - 46202994 [-] on Build GRCh38 72
          GenBankmRNA DW413615.1 - 1 - 2 262 Chr. 19 - 46198385 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW419174.1 - 1 - 2 246 Chr. 19 - 46198281 - 46203067 [-] on Build GRCh38 72
          GenBankmRNA DW452244.1 - 1 - 2 262 Chr. 19 - 46198334 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW465475.1 - 1 - 2 262 Chr. 19 - 46195778 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW452041.1 - 1 - 2 262 Chr. 19 - 46198332 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW407902.1 - 1 - 2 Chr. 19 - 46198285 - 46203083 [+] on Build GRCh38 72
          GenBankmRNA DW418034.1 - 1 - 2 261 Chr. 19 - 46198399 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW463158.1 - 1 - 2 262 Chr. 19 - 46198341 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA BM562975.1 - 1 - 2 224 Chr. 19 - 46189645 - 46203062 [-] on Build GRCh38 72
          GenBankmRNA DW466981.1 - 1 - 2 169 Chr. 19 - 46198262 - 46202991 [-] on Build GRCh38 72
          GenBankmRNA DW420827.1 - 1 - 2 262 Chr. 19 - 46198405 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW415880.1 - 1 - 2 213 Chr. 19 - 46189166 - 46203034 [-] on Build GRCh38 72
          GenBankmRNA DW462337.1 - 1 - 2 262 Chr. 19 - 46198335 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW419128.1 - 1 - 2 262 Chr. 19 - 46198296 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW419268.1 - 1 - 2 262 Chr. 19 - 46198299 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW408868.1 - 1 - 2 262 Chr. 19 - 46198363 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW408083.1 - 1 - 2 262 Chr. 19 - 46198285 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW416702.1 - 1 - 2 262 Chr. 19 - 46198396 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW415827.1 - 1 - 2 262 Chr. 19 - 46198393 - 46203083 [-] on Build GRCh38 72
          GenBankmRNA DW408687.1 - 1 - 2 262 Chr. 19 - 46198362 - 46203083 [-] on Build GRCh38 72

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream IGF like family member 2 IGFL2 UNQ645,
          VPRI645
          -3.3kb Anti-sense Chr. 19 - 46078513 - 46195097 on NCBI Build GRCh38
          Downstream uncharacterized LOC93429 LOC93429 7.2kb Anti-sense Chr. 19 - 46210242 - 46214838 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Homo sapiens Probe spans exons

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000594027.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000594027.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000594027.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000594027.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000594027.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000594027.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000594027.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000594027.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000594027.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000594027.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000595673.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000595673.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000595673.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000595673.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000595673.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000598518.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000598518.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000598518.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000598518.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000598518.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000598518.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000598518.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000598518.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000598518.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000598518.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000598518.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000598518.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000598754.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000598754.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000598754.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000598754.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000598754.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000599817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000599817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000599817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000599817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000599817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000599817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000599817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000599817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000599817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000599817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000599817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000599817.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000601263.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000601263.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000601263.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010974.hg.1 ENST00000601263.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000594027.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000594027.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000594027.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000594027.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000594027.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000594027.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000594027.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000594027.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000594027.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000594027.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000595673.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000595673.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000595673.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000595673.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000595673.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000598518.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000598518.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000598518.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000598518.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000598518.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000598518.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000598518.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000598518.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000598518.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000598518.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000598518.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000598518.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000598754.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000598754.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000598754.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000598754.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000598754.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000599817.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000599817.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000599817.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000599817.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000599817.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000599817.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000599817.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000599817.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000599817.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000599817.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000599817.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000599817.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000601263.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000601263.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000601263.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010974.hg.1 ENST00000601263.1 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0