Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105371814 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_135674.1 | 3 - 4 | 561 | 85 | ||
GenBank mRNA | CX873301.1 | - | 3 - 4 | 564 | 85 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BF244886.1 | - | 145 | Chr. - - [-] on Build | 84 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | calcium binding and coiled-coil domain 2 | CALCOCO2 | NDP52 | -9.6kb | Anti-sense | Chr. 17 - 48830988 - 48865245 on NCBI Build GRCh38 |
Downstream |
ATP synthase, H+ transporting, mitochondrial Fo complex subunit C1 (subunit 9) |
ATP5G1 |
ATP5A, ATP5G |
13.4kb | Anti-sense | Chr. 17 - 48892786 - 48895871 on NCBI Build GRCh38 |
Set Membership: |
![]() ![]() ![]() ![]() |