Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010647.hg.1 |
ENST00000560068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010647.hg.1 |
ENST00000560068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010647.hg.1 |
ENST00000560068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010647.hg.1 |
ENST00000560068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010647.hg.1 |
ENST00000560068.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010647.hg.1 |
ENST00000560351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010647.hg.1 |
ENST00000560351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010647.hg.1 |
ENST00000560351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010647.hg.1 |
ENST00000560351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010647.hg.1 |
ENST00000560351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010647.hg.1 |
ENST00000560351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010647.hg.1 |
ENST00000560351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010647.hg.1 |
ENST00000560351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010647.hg.1 |
ENST00000560351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010647.hg.1 |
ENST00000560461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010647.hg.1 |
ENST00000560461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010647.hg.1 |
ENST00000560461.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010647.hg.1 |
ENST00000560461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010647.hg.1 |
ENST00000560068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010647.hg.1 |
ENST00000560068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010647.hg.1 |
ENST00000560068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010647.hg.1 |
ENST00000560068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010647.hg.1 |
ENST00000560068.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010647.hg.1 |
ENST00000560351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010647.hg.1 |
ENST00000560351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010647.hg.1 |
ENST00000560351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010647.hg.1 |
ENST00000560351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010647.hg.1 |
ENST00000560351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010647.hg.1 |
ENST00000560351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010647.hg.1 |
ENST00000560351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010647.hg.1 |
ENST00000560351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010647.hg.1 |
ENST00000560351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010647.hg.1 |
ENST00000560461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010647.hg.1 |
ENST00000560461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010647.hg.1 |
ENST00000560461.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010647.hg.1 |
ENST00000560461.1 |
Ensembl |