Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006733.hg.1 |
ENST00000424453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006733.hg.1 |
ENST00000424453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006733.hg.1 |
ENST00000424453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006733.hg.1 |
ENST00000424453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006733.hg.1 |
ENST00000443874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006733.hg.1 |
ENST00000443874.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006733.hg.1 |
ENST00000443874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006733.hg.1 |
ENST00000418428.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006733.hg.1 |
ENST00000424453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006733.hg.1 |
ENST00000424453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006733.hg.1 |
ENST00000424453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006733.hg.1 |
ENST00000424453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006733.hg.1 |
ENST00000443874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006733.hg.1 |
ENST00000443874.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006733.hg.1 |
ENST00000443874.1 |
Ensembl |