Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009695.hg.1 |
ENST00000448761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009695.hg.1 |
ENST00000448761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009695.hg.1 |
ENST00000448761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009695.hg.1 |
ENST00000448761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009695.hg.1 |
ENST00000448761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009695.hg.1 |
NR_110654.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009695.hg.1 |
NR_110654.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009695.hg.1 |
NR_110654.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009695.hg.1 |
NR_110654.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009695.hg.1 |
NR_110654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009695.hg.1 |
ENST00000448761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009695.hg.1 |
ENST00000448761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009695.hg.1 |
ENST00000448761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009695.hg.1 |
ENST00000448761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009695.hg.1 |
ENST00000448761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009695.hg.1 |
NR_110654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009695.hg.1 |
NR_110654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009695.hg.1 |
NR_110654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009695.hg.1 |
NR_110654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009695.hg.1 |
NR_110654.1 |
Refseq |