Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927348 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110915.1 | 1 - 2 | 139 | 136 | ||
GenBank mRNA | BC039386.1 | - | 1 - 2 | 139 | 136 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101927311 | LOC101927311 | -4.3kb | Sense | Chr. 16 - 14009276 - 14016018 on NCBI Build GRCh38 | |
Downstream | MKL1/myocardin like 2 | MKL2 |
MRTF-B, MRTFB, NPD001 |
50.0kb | Anti-sense | Chr. 16 - 14071076 - 14266779 on NCBI Build GRCh38 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |