Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs04972233_s1
See other SF1 GE Assays ›
Gene Symbol:
SF1
Gene Name:
splicing factor 1
Gene Aliases:
BBP, D11S636, MBBP, ZCCHC25, ZFM1, ZNF162
Chromosome Location:
Chr.11: 64764604 - 64779043 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs04972233_s1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: SF1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001178030.1
NP_001171501.1
13
2475
106
NM_001178031.1
NP_001171502.1
13
2894
106
NM_004630.3
NP_004621.2
13
3115
106
NM_201995.2
NP_973724.1
14
2683
106
NM_201997.2
NP_973726.2
13
2482
106
NM_201998.2
NP_973727.1
13
2478
106
NR_033649.1
12
2402
106
NR_033650.1
12
2406
106
XM_011545244.1
XP_011543546.1
14
2355
106
XM_011545245.1
XP_011543547.1
13
2791
106
XM_011545246.1
XP_011543548.1
13
2782
106
XM_011545247.1
XP_011543549.1
14
2359
106
XM_011545248.1
XP_011543550.1
13
2154
106
XM_017018244.1
XP_016873733.1
13
2787
106
XM_017018245.1
XP_016873734.1
13
2868
106
XM_017018246.1
XP_016873735.1
13
2145
106
XM_017018247.1
XP_016873736.1
13
2825
106
XM_017018248.1
XP_016873737.1
13
2188
106
XM_017018249.1
XP_016873738.1
13
5280
106
XM_017018250.1
XP_016873739.1
12
2964
106
XM_017018251.1
XP_016873740.1
12
2819
106
GenBank mRNA
BC000773.1
-
4
1543
106
BC008080.2
-
13
2174
2961666
106
BC008724.2
-
13
2174
2961666
106
BC020217.1
-
13
2377
4869292
106
BC034451.1
-
14
2608
106
BC038446.1
-
13
2498
5493676
106
BQ230035.1
-
3
610
106
BU624935.1
-
1
106
D26120.1
-
14
2662
106
L49345.1
-
13
2368
106
L49380.1
-
13
3001
106
Y08765.1
-
13
2705
106
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length greater than or equal to 101
3' Most
Mammalian Gene Collection
Detect Genomic DNA
Homo sapiens
Both primers and probe map within a single exon
Matching Ambion siRNAs:
Click here to find siRNAs for Gene SF1
Gene Ontology Categories:
Function(s)
Process(es)
spliceosomal complex assembly
mRNA 3'-splice site recognition
mRNA splicing, via spliceosome
transcription, DNA-templated
regulation of transcription, DNA-templated
male sex determination
Leydig cell differentiation
negative regulation of smooth muscle cell proliferation
regulation of steroid biosynthetic process
negative regulation of nucleic acid-templated transcription
transcription corepressor activity
RNA binding
protein binding
zinc ion binding
identical protein binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0