Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009944.hg.1 |
ENST00000329759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009944.hg.1 |
ENST00000329759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009944.hg.1 |
ENST00000329759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009944.hg.1 |
ENST00000329759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009944.hg.1 |
ENST00000329759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009944.hg.1 |
ENST00000329759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009944.hg.1 |
NM_153690.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009944.hg.1 |
NM_153690.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009944.hg.1 |
NM_153690.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009944.hg.1 |
NM_153690.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009944.hg.1 |
NM_153690.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009944.hg.1 |
NM_153690.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009944.hg.1 |
NM_153690.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009944.hg.1 |
NM_153690.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009944.hg.1 |
NM_153690.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009944.hg.1 |
ENST00000329759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009944.hg.1 |
ENST00000329759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009944.hg.1 |
ENST00000329759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009944.hg.1 |
ENST00000329759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009944.hg.1 |
ENST00000329759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009944.hg.1 |
ENST00000329759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009944.hg.1 |
NM_153690.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009944.hg.1 |
NM_153690.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009944.hg.1 |
NM_153690.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009944.hg.1 |
NM_153690.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009944.hg.1 |
NM_153690.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009944.hg.1 |
NM_153690.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009944.hg.1 |
NM_153690.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009944.hg.1 |
NM_153690.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009944.hg.1 |
NM_153690.4 |
Refseq |