Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
ENST00000304771.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
ENST00000304771.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
ENST00000304771.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
ENST00000304771.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
ENST00000304771.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
ENST00000304771.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
ENST00000304771.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
ENST00000304771.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
ENST00000304771.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
ENST00000304771.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
ENST00000304771.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
ENST00000304771.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
ENST00000304771.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
ENST00000434603.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
ENST00000434603.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
ENST00000434603.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
ENST00000434603.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
ENST00000434603.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
ENST00000434603.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
ENST00000434603.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
ENST00000526689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
ENST00000526689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
ENST00000526740.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
ENST00000526740.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
ENST00000526740.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
ENST00000526740.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
ENST00000526740.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
ENST00000526740.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
ENST00000526740.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
NM_033063.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
NM_033063.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
NM_033063.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
NM_033063.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
NM_033063.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
NM_033063.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
NM_033063.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
NM_033063.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
NM_033063.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
NM_033063.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
NM_207577.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
NM_207577.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
NM_207577.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
NM_207577.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
NM_207577.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
NM_207577.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011658.hg.1 |
NM_207577.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011660.hg.1 |
ENST00000530435.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011660.hg.1 |
ENST00000530435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011660.hg.1 |
ENST00000530435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011660.hg.1 |
ENST00000530435.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
ENST00000304771.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
ENST00000304771.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
ENST00000304771.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
ENST00000304771.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
ENST00000304771.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
ENST00000304771.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
ENST00000304771.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
ENST00000304771.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
ENST00000304771.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
ENST00000304771.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
ENST00000304771.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
ENST00000304771.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
ENST00000304771.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
ENST00000434603.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
ENST00000434603.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
ENST00000434603.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
ENST00000434603.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
ENST00000434603.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
ENST00000434603.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
ENST00000434603.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
ENST00000526689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
ENST00000526689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
ENST00000526740.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
ENST00000526740.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
ENST00000526740.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
ENST00000526740.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
ENST00000526740.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
ENST00000526740.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
ENST00000526740.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
NM_033063.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
NM_033063.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
NM_033063.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
NM_033063.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
NM_033063.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
NM_033063.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
NM_033063.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
NM_033063.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
NM_033063.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
NM_033063.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
NM_207577.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
NM_207577.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
NM_207577.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
NM_207577.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
NM_207577.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
NM_207577.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011658.hg.1 |
NM_207577.1 |
Refseq |