Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
ENST00000455399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
ENST00000455399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
ENST00000455399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
ENST00000455399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
ENST00000455399.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009233.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
ENST00000455399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
ENST00000455399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
ENST00000455399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
ENST00000455399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
ENST00000455399.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
ENST00000608254.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
NR_073010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009233.hg.1 |
NR_073010.2 |
Refseq |