Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs05001831_m1
See other ANK3 GE Assays ›
Gene Symbol:
ANK3
Gene Name:
ankyrin 3, node of Ranvier (ankyrin G)
Gene Aliases:
ANKYRIN-G, MRT37
Chromosome Location:
Chr.10: 60026298 - 60733526 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs05001831_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ANK3
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001204404.1
NP_001191333.1
24 - 25
3049
127
XM_005269715.3
XP_005269772.1
24 - 25
2950
127
XM_006717796.3
XP_006717859.1
24 - 25
2950
127
XM_006717802.3
XP_006717865.1
24 - 25
2950
127
XM_011539708.2
XP_011538010.1
24 - 25
2950
127
XM_011539709.2
XP_011538011.1
24 - 25
2950
127
XM_017016110.1
XP_016871599.1
24 - 25
2950
127
XM_017016111.1
XP_016871600.1
24 - 25
2950
127
XM_017016112.1
XP_016871601.1
24 - 25
2950
127
XM_017016113.1
XP_016871602.1
24 - 25
2950
127
XM_017016114.1
XP_016871603.1
24 - 25
3049
127
XM_017016115.1
XP_016871604.1
24 - 25
2779
127
XM_017016116.1
XP_016871605.1
24 - 25
2950
127
XM_017016117.1
XP_016871606.1
24 - 25
2950
127
XM_017016118.1
XP_016871607.1
24 - 25
2950
127
XM_017016119.1
XP_016871608.1
24 - 25
2950
127
XM_017016120.1
XP_016871609.1
24 - 25
2950
127
XM_017016121.1
XP_016871610.1
24 - 25
2950
127
XM_017016123.1
XP_016871612.1
24 - 25
2950
127
XM_017016124.1
XP_016871613.1
24 - 25
2950
127
XM_017016125.1
XP_016871614.1
24 - 25
2950
127
XM_017016128.1
XP_016871617.1
24 - 25
2950
127
XM_017016129.1
XP_016871618.1
24 - 25
2950
127
XM_017016136.1
XP_016871625.1
24 - 25
2950
127
XM_017016137.1
XP_016871626.1
24 - 25
2950
127
XM_017016138.1
XP_016871627.1
24 - 25
2950
127
GenBank mRNA
AK295661.1
-
24 - 25
2756
127
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
Homo sapiens
Probe spans exons
Gene Ontology Categories:
Function(s)
Process(es)
mitotic cytokinesis
endoplasmic reticulum to Golgi vesicle-mediated transport
plasma membrane organization
signal transduction
axonogenesis
axon guidance
neuromuscular junction development
positive regulation of gene expression
positive regulation of cell communication by electrical coupling
positive regulation of sodium ion transport
magnesium ion homeostasis
neuronal action potential
positive regulation of homotypic cell-cell adhesion
Golgi to plasma membrane protein transport
regulation of potassium ion transport
establishment of protein localization
positive regulation of membrane potential
cellular response to magnesium ion
membrane assembly
protein localization to plasma membrane
maintenance of protein location in plasma membrane
positive regulation of protein targeting to membrane
positive regulation of membrane depolarization during cardiac muscle cell action potential
negative regulation of delayed rectifier potassium channel activity
positive regulation of sodium ion transmembrane transporter activity
positive regulation of cation channel activity
structural constituent of cytoskeleton
protein binding
cytoskeletal protein binding
cytoskeletal anchor activity
spectrin binding
protein-macromolecule adaptor activity
transmembrane transporter binding
cadherin binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0