Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007506.hg.1 |
ENST00000255152.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007506.hg.1 |
ENST00000255152.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007506.hg.1 |
ENST00000255152.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007506.hg.1 |
ENST00000255152.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007506.hg.1 |
NM_080752.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007506.hg.1 |
NM_080752.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007506.hg.1 |
NM_080752.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007506.hg.1 |
NM_080752.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007506.hg.1 |
NM_080752.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007506.hg.1 |
NR_037628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007506.hg.1 |
NR_037628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007506.hg.1 |
NR_037628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007506.hg.1 |
NR_037628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007506.hg.1 |
NR_037628.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007506.hg.1 |
NR_037628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007506.hg.1 |
ENST00000255152.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007506.hg.1 |
ENST00000255152.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007506.hg.1 |
ENST00000255152.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007506.hg.1 |
ENST00000255152.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007506.hg.1 |
NM_080752.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007506.hg.1 |
NM_080752.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007506.hg.1 |
NM_080752.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007506.hg.1 |
NM_080752.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007506.hg.1 |
NM_080752.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007506.hg.1 |
NR_037628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007506.hg.1 |
NR_037628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007506.hg.1 |
NR_037628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007506.hg.1 |
NR_037628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007506.hg.1 |
NR_037628.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007506.hg.1 |
NR_037628.1 |
Refseq |