Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05029754_s1
          See other SOX4 GE Assays ›
          Gene Symbol:
          SOX4
          Gene Name:
          SRY-box 4
          Gene Aliases:
          EVI16
          Chromosome Location:
          Chr.6: 21593741 - 21598619 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05029754_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SOX4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003107.2 NP_003098.1 1 799 80
          GenBank mRNA BC072668.1 - 2 619 6584346 80
          X70683.1 - 1 355 80

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the BC072668.1, NM_003107.2, X70683.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SOX4

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          neural tube formation
          pro-B cell differentiation
          mitral valve morphogenesis
          cardiac ventricle formation
          cardiac right ventricle morphogenesis
          atrial septum primum morphogenesis
          noradrenergic neuron differentiation
          regulation of transcription, DNA-templated
          transcription by RNA polymerase II
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          heart development
          positive regulation of cell population proliferation
          negative regulation of cell population proliferation
          glial cell proliferation
          spinal cord development
          spinal cord motor neuron differentiation
          glial cell development
          positive regulation of Wnt signaling pathway
          T cell differentiation
          endocrine pancreas development
          negative regulation of protein ubiquitination
          regulation of protein stability
          positive regulation of insulin secretion
          somatic stem cell population maintenance
          ascending aorta morphogenesis
          glucose homeostasis
          DNA damage response, detection of DNA damage
          positive regulation of apoptotic process
          positive regulation of translation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          negative regulation of protein export from nucleus
          sympathetic nervous system development
          protein stabilization
          canonical Wnt signaling pathway
          limb bud formation
          ventricular septum morphogenesis
          negative regulation of cell death
          neuroepithelial cell differentiation
          kidney morphogenesis
          cellular response to glucose stimulus
          positive regulation of canonical Wnt signaling pathway
          positive regulation of N-terminal peptidyl-lysine acetylation
          transcription cis-regulatory region binding
          core promoter sequence-specific DNA binding
          DNA-binding transcription factor activity
          transcription coactivator activity
          protein binding
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0600007158.hg.1 ENST00000244745.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007158.hg.1 ENST00000244745.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007158.hg.1 ENST00000244745.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007158.hg.1 ENST00000244745.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007158.hg.1 ENST00000244745.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007158.hg.1 ENST00000244745.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007158.hg.1 ENST00000244745.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007158.hg.1 ENST00000244745.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007158.hg.1 ENST00000244745.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007158.hg.1 ENST00000244745.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007158.hg.1 ENST00000244745.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007158.hg.1 NM_003107.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007158.hg.1 NM_003107.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007158.hg.1 NM_003107.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007158.hg.1 NM_003107.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007158.hg.1 NM_003107.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007158.hg.1 NM_003107.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007158.hg.1 NM_003107.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007158.hg.1 NM_003107.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007158.hg.1 NM_003107.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007158.hg.1 NM_003107.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007158.hg.1 ENST00000244745.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600007158.hg.1 ENST00000244745.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600007158.hg.1 ENST00000244745.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600007158.hg.1 ENST00000244745.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600007158.hg.1 ENST00000244745.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600007158.hg.1 ENST00000244745.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600007158.hg.1 ENST00000244745.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600007158.hg.1 ENST00000244745.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600007158.hg.1 ENST00000244745.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600007158.hg.1 ENST00000244745.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600007158.hg.1 ENST00000244745.2 Ensembl
          Affymetrix Human Clariom D Assay TC0600007158.hg.1 NM_003107.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007158.hg.1 NM_003107.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007158.hg.1 NM_003107.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007158.hg.1 NM_003107.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007158.hg.1 NM_003107.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007158.hg.1 NM_003107.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007158.hg.1 NM_003107.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007158.hg.1 NM_003107.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007158.hg.1 NM_003107.2 Refseq
          Affymetrix Human Clariom D Assay TC0600007158.hg.1 NM_003107.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0