Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011179.hg.1 |
ENST00000328087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011179.hg.1 |
ENST00000328087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011179.hg.1 |
ENST00000328087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011179.hg.1 |
ENST00000328087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011179.hg.1 |
ENST00000328087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011179.hg.1 |
ENST00000328087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011179.hg.1 |
ENST00000328087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011179.hg.1 |
ENST00000328087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011179.hg.1 |
ENST00000328087.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011179.hg.1 |
ENST00000538666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011179.hg.1 |
ENST00000538666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011179.hg.1 |
ENST00000538666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011179.hg.1 |
ENST00000538666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011179.hg.1 |
ENST00000538666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011179.hg.1 |
ENST00000538666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011179.hg.1 |
ENST00000538666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011179.hg.1 |
ENST00000538666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011179.hg.1 |
ENST00000538666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011179.hg.1 |
NM_020525.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011179.hg.1 |
NM_020525.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011179.hg.1 |
NM_020525.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011179.hg.1 |
NM_020525.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011179.hg.1 |
NM_020525.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011179.hg.1 |
NM_020525.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011179.hg.1 |
NM_020525.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011179.hg.1 |
NM_020525.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011179.hg.1 |
NM_020525.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011179.hg.1 |
ENST00000328087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011179.hg.1 |
ENST00000328087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011179.hg.1 |
ENST00000328087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011179.hg.1 |
ENST00000328087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011179.hg.1 |
ENST00000328087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011179.hg.1 |
ENST00000328087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011179.hg.1 |
ENST00000328087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011179.hg.1 |
ENST00000328087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011179.hg.1 |
ENST00000328087.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011179.hg.1 |
ENST00000538666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011179.hg.1 |
ENST00000538666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011179.hg.1 |
ENST00000538666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011179.hg.1 |
ENST00000538666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011179.hg.1 |
ENST00000538666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011179.hg.1 |
ENST00000538666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011179.hg.1 |
ENST00000538666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011179.hg.1 |
ENST00000538666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011179.hg.1 |
ENST00000538666.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011179.hg.1 |
NM_020525.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011179.hg.1 |
NM_020525.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011179.hg.1 |
NM_020525.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011179.hg.1 |
NM_020525.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011179.hg.1 |
NM_020525.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011179.hg.1 |
NM_020525.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011179.hg.1 |
NM_020525.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011179.hg.1 |
NM_020525.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011179.hg.1 |
NM_020525.4 |
Refseq |