Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05035700_s1
          See other PSMB5 GE Assays ›
          Gene Symbol:
          PSMB5
          Gene Name:
          proteasome subunit beta 5
          Gene Aliases:
          LMPX, MB1, X
          Chromosome Location:
          Chr.14: 23016543 - 23035220 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05035700_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PSMB5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001144932.2 NP_001138404.1 1 370 101
          NM_002797.4 NP_002788.1 1 370 101
          XM_005267871.2 XP_005267928.1 1 365 101
          GenBank mRNA AK225888.1 - 1 135 101
          AK300714.1 - 1 115 101
          AK311895.1 - 1 126 101
          BC057840.1 - 1 125 4795732 101
          BC107720.1 - 1 121 6738409 101
          BX248299.1 - 1 102 101
          BX538001.1 - 1 117 101

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PSMB5

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          protein polyubiquitination
          stimulatory C-type lectin receptor signaling pathway
          antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
          proteolysis
          regulation of cellular amino acid metabolic process
          response to oxidative stress
          viral process
          anaphase-promoting complex-dependent catabolic process
          tumor necrosis factor-mediated signaling pathway
          NIK/NF-kappaB signaling
          Fc-epsilon receptor signaling pathway
          proteasome-mediated ubiquitin-dependent protein catabolic process
          regulation of mRNA stability
          T cell receptor signaling pathway
          Wnt signaling pathway, planar cell polarity pathway
          negative regulation of canonical Wnt signaling pathway
          positive regulation of canonical Wnt signaling pathway
          threonine-type endopeptidase activity
          protein binding
          peptidase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000334454.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000334454.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000334454.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000334454.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000334454.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000334454.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000334454.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000334454.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000361611.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000361611.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000361611.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000361611.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000361611.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000361611.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000361611.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000361611.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000361611.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000361611.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000361611.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000361611.10 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000385031.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000385031.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000425762.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000425762.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000425762.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000425762.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000425762.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000425762.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000425762.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000425762.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000425762.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000460922.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000460922.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000460922.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000460922.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000460922.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000460922.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000493471.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000493471.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000493471.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000493471.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000493471.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000493471.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000493471.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000493471.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000493471.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000493471.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000493471.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000555895.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000555895.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000555895.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000555895.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000555895.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 ENST00000555895.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_001130725.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_001130725.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_001130725.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_001130725.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_001130725.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_001130725.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_001130725.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_001130725.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_001130725.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_001130725.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_001144932.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_001144932.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_001144932.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_001144932.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_001144932.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_001144932.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_001144932.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_001144932.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_001144932.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_001144932.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_001144932.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_001144932.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_001144932.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_002797.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_002797.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_002797.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_002797.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_002797.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_002797.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_002797.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_002797.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_002797.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_002797.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_002797.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1400008689.hg.1 NM_002797.4 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000334454.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000334454.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000334454.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000334454.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000334454.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000334454.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000334454.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000334454.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000361611.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000361611.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000361611.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000361611.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000361611.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000361611.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000361611.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000361611.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000361611.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000361611.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000361611.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000361611.10 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000385031.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000385031.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000425762.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000425762.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000425762.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000425762.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000425762.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000425762.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000425762.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000425762.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000425762.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000460922.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000460922.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000460922.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000460922.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000460922.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000460922.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000493471.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000493471.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000493471.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000493471.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000493471.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000493471.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000493471.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000493471.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000493471.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000493471.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000493471.2 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000555895.5 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000555895.5 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000555895.5 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000555895.5 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000555895.5 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 ENST00000555895.5 Ensembl
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_001130725.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_001130725.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_001130725.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_001130725.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_001130725.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_001130725.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_001130725.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_001130725.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_001130725.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_001130725.1 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_001144932.2 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_001144932.2 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_001144932.2 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_001144932.2 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_001144932.2 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_001144932.2 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_001144932.2 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_001144932.2 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_001144932.2 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_001144932.2 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_001144932.2 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_001144932.2 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_001144932.2 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_002797.4 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_002797.4 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_002797.4 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_002797.4 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_002797.4 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_002797.4 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_002797.4 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_002797.4 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_002797.4 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_002797.4 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_002797.4 Refseq
          Affymetrix Human Clariom D Assay TC1400008689.hg.1 NM_002797.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix