Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008262.hg.1 |
ENST00000301838.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008262.hg.1 |
ENST00000301838.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008262.hg.1 |
ENST00000301838.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008262.hg.1 |
ENST00000301838.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008262.hg.1 |
ENST00000301838.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008262.hg.1 |
NM_003824.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008262.hg.1 |
NM_003824.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008262.hg.1 |
NM_003824.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008262.hg.1 |
NM_003824.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008262.hg.1 |
NM_003824.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011470.hg.1 |
ENST00000526174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011470.hg.1 |
ENST00000526174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011470.hg.1 |
ENST00000526174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011470.hg.1 |
ENST00000527232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011470.hg.1 |
ENST00000527232.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011470.hg.1 |
ENST00000527232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008262.hg.1 |
ENST00000301838.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008262.hg.1 |
ENST00000301838.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008262.hg.1 |
ENST00000301838.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008262.hg.1 |
ENST00000301838.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008262.hg.1 |
ENST00000301838.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008262.hg.1 |
NM_003824.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008262.hg.1 |
NM_003824.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008262.hg.1 |
NM_003824.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008262.hg.1 |
NM_003824.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008262.hg.1 |
NM_003824.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011470.hg.1 |
ENST00000526174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011470.hg.1 |
ENST00000526174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011470.hg.1 |
ENST00000526174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011470.hg.1 |
ENST00000527232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011470.hg.1 |
ENST00000527232.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011470.hg.1 |
ENST00000527232.1 |
Ensembl |