Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006686.hg.1 |
ENST00000506655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006686.hg.1 |
ENST00000506655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006686.hg.1 |
ENST00000510067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006686.hg.1 |
ENST00000510067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006686.hg.1 |
ENST00000510067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006686.hg.1 |
ENST00000510067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006686.hg.1 |
ENST00000510067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006686.hg.1 |
ENST00000510067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006686.hg.1 |
ENST00000510067.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006686.hg.1 |
ENST00000510229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006686.hg.1 |
ENST00000510229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006686.hg.1 |
ENST00000510229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006686.hg.1 |
ENST00000510229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006686.hg.1 |
NR_109932.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006686.hg.1 |
NR_109932.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006686.hg.1 |
NR_109932.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006686.hg.1 |
NR_109932.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500006686.hg.1 |
NR_109932.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006686.hg.1 |
ENST00000506655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006686.hg.1 |
ENST00000506655.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006686.hg.1 |
ENST00000510067.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006686.hg.1 |
ENST00000510067.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006686.hg.1 |
ENST00000510067.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006686.hg.1 |
ENST00000510067.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006686.hg.1 |
ENST00000510067.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006686.hg.1 |
ENST00000510067.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006686.hg.1 |
ENST00000510067.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006686.hg.1 |
ENST00000510229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006686.hg.1 |
ENST00000510229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006686.hg.1 |
ENST00000510229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006686.hg.1 |
ENST00000510229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500006686.hg.1 |
NR_109932.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006686.hg.1 |
NR_109932.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006686.hg.1 |
NR_109932.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006686.hg.1 |
NR_109932.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500006686.hg.1 |
NR_109932.1 |
Refseq |