Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05049750_s1
          See other TBATA GE Assays ›
          Gene Symbol:
          TBATA
          Gene Name:
          thymus, brain and testes associated
          Gene Aliases:
          C10orf27, SPATIAL
          Chromosome Location:
          Chr.10: 70771237 - 70785419 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05049750_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TBATA
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001318241.1 NP_001305170.1 11 1567 64
          NM_001318242.1 NP_001305171.1 11 1631 64
          NM_001318243.1 NP_001305172.1 11 1561 64
          NM_152710.3 NP_689923.3 11 1564 64
          NR_134531.1 10 1482 64
          NR_134532.1 11 1490 64
          NR_134533.1 7 1100 64
          NR_134534.1 6 1024 64
          XM_017015845.1 XP_016871334.1 13 2143 64
          XM_017015846.1 XP_016871335.1 13 2027 64
          XM_017015847.1 XP_016871336.1 12 1890 64
          XM_017015848.1 XP_016871337.1 12 1691 64
          XM_017015852.1 XP_016871341.1 11 1589 64
          XM_017015853.1 XP_016871342.1 12 1642 64
          XM_017015856.1 XP_016871345.1 12 1795 64
          XM_017015860.1 XP_016871349.1 10 1415 64
          GenBank mRNA AK057382.1 - 11 1546 64
          AK124147.1 - 1 2691 64
          DB512647.2 - 6 673 64
          HY289269.1 - 1 64

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TBATA

          Gene Ontology Categories:

          Function(s)

          multicellular organism development
          spermatogenesis
          cell differentiation

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000299290.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000299290.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000299290.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000299290.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000299290.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000299290.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000299290.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000299290.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000299290.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000299290.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000299290.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000299290.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000299290.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000299290.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000299290.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000394982.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000394982.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000394982.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000394982.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000394982.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000456372.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000456372.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000456372.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000456372.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000456372.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000456372.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000456372.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000456372.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000456372.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000456372.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000456372.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000456372.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000456372.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000456372.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 ENST00000456372.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 NM_152710.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 NM_152710.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 NM_152710.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 NM_152710.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 NM_152710.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 NM_152710.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 NM_152710.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 NM_152710.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 NM_152710.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 NM_152710.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 NM_152710.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 NM_152710.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 NM_152710.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 NM_152710.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010926.hg.1 NM_152710.3 Refseq
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000299290.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000299290.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000299290.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000299290.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000299290.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000299290.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000299290.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000299290.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000299290.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000299290.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000299290.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000299290.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000299290.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000299290.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000299290.5 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000394982.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000394982.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000394982.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000394982.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000394982.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000456372.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000456372.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000456372.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000456372.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000456372.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000456372.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000456372.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000456372.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000456372.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000456372.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000456372.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000456372.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000456372.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000456372.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 ENST00000456372.3 Ensembl
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 NM_152710.3 Refseq
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 NM_152710.3 Refseq
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 NM_152710.3 Refseq
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 NM_152710.3 Refseq
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 NM_152710.3 Refseq
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 NM_152710.3 Refseq
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 NM_152710.3 Refseq
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 NM_152710.3 Refseq
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 NM_152710.3 Refseq
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 NM_152710.3 Refseq
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 NM_152710.3 Refseq
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 NM_152710.3 Refseq
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 NM_152710.3 Refseq
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 NM_152710.3 Refseq
          Affymetrix Human Clariom D Assay TC1000010926.hg.1 NM_152710.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-867f8cf58d-sfwzd:80/100.66.130.62:80.
          git-commit: 7a7e1ef32845d1da800a461bcf18a29ea3e3afc0
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix