Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014222.hg.1 |
ENST00000604204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014222.hg.1 |
ENST00000604204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014222.hg.1 |
NR_033884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014222.hg.1 |
NR_033884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014222.hg.1 |
NR_033884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014222.hg.1 |
NR_033884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014222.hg.1 |
NR_033884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014222.hg.1 |
NR_033884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014222.hg.1 |
NR_033884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014222.hg.1 |
NR_033884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014222.hg.1 |
NR_033884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014222.hg.1 |
NR_033884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014222.hg.1 |
NR_033884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014222.hg.1 |
ENST00000604204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014222.hg.1 |
ENST00000604204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014222.hg.1 |
NR_033884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014222.hg.1 |
NR_033884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014222.hg.1 |
NR_033884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014222.hg.1 |
NR_033884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014222.hg.1 |
NR_033884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014222.hg.1 |
NR_033884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014222.hg.1 |
NR_033884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014222.hg.1 |
NR_033884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014222.hg.1 |
NR_033884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014222.hg.1 |
NR_033884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014222.hg.1 |
NR_033884.1 |
Refseq |