Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05056709_g1
          See other PLEKHA6 GE Assays ›
          Gene Symbol:
          PLEKHA6
          Gene Name:
          pleckstrin homology domain containing A6
          Gene Aliases:
          PEPP-3, PEPP3
          Chromosome Location:
          Chr.1: 204218851 - 204378376 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05056709_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PLEKHA6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_014935.4 NP_055750.2 9 1748 59
          XM_005244966.4 XP_005245023.2 9 1615 59
          XM_005244967.4 XP_005245024.2 9 1674 59
          XM_005244968.4 XP_005245025.2 9 1614 59
          XM_006711212.3 XP_006711275.1 9 1901 59
          XM_006711213.3 XP_006711276.2 9 1675 59
          XM_006711214.3 XP_006711277.2 9 1675 59
          XM_006711215.3 XP_006711278.2 9 1675 59
          XM_006711217.3 XP_006711280.2 9 1675 59
          XM_006711219.3 XP_006711282.2 9 1675 59
          XM_006711221.2 XP_006711284.1 9 1999 59
          XM_006711222.3 XP_006711285.1 9 1984 59
          XM_011509295.2 XP_011507597.1 9 1675 59
          XM_011509296.2 XP_011507598.1 9 1675 59
          XM_011509297.2 XP_011507599.1 11 2501 59
          XM_011509298.2 XP_011507600.1 9 1784 59
          XM_011509299.2 XP_011507601.2 9 1790 59
          XM_011509301.2 XP_011507603.1 9 5680 59
          XM_011509304.2 XP_011507606.1 9 1629 59
          XM_017000687.1 XP_016856176.1 9 1675 59
          XM_017000688.1 XP_016856177.1 9 1675 59
          XM_017000689.1 XP_016856178.1 9 1615 59
          XM_017000690.1 XP_016856179.1 9 5620 59
          XM_017000691.1 XP_016856180.1 4 4073 59
          XM_017000692.1 XP_016856181.1 4 4013 59
          GenBank mRNA AB023186.1 - 9 1735 59
          BC152475.1 - 9 1735 8862612 59
          DQ925685.1 - 1 21 59
          DQ925686.1 - 1 21 59
          DQ925687.1 - 1 21 59
          DQ925688.1 - 1 21 59
          DQ925689.1 - 1 21 59
          DQ925690.1 - 1 21 59
          DQ925691.1 - 1 21 59

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PLEKHA6

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000485632.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000485632.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 ENST00000485632.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000272203.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000414478.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000485632.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000485632.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 ENST00000485632.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 NM_014935.4 Refseq
          Affymetrix Human Clariom D Assay TC0100018553.hg.1 NM_014935.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0