Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05362169_g1
          See other WASH1 GE Assays ›
          Gene Symbol:
          WASH1 WASH5P
          Gene Name:
          WAS protein family homolog 1
          WAS protein family homolog 5 pseudogene
          Gene Aliases:
          FAM39E, WASH
          -
          Chromosome Location:
          Chr.9: 14475 - 30487 on Build GRCh38
          Chromosome Location:
          Chr.19: 60951 - 70966 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs05362169_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: WASH1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_011517663.2 XP_011515965.1 1 346 92
          XM_011517664.2 XP_011515966.1 1 346 92
          Gene Symbol: WASH5P
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033266.1 3 359 92
          GenBank mRNA AK308635.1 - 3 369 92

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the XM_011517663.2, XM_011517664.2 transcripts.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          exocytosis
          protein transport
          endosomal transport
          extracellular matrix disassembly
          positive regulation of cell migration
          positive regulation of pseudopodium assembly
          Arp2/3 complex-mediated actin nucleation
          retrograde transport, endosome to Golgi
          retrograde transport, endosome to plasma membrane
          actin binding
          protein binding
          ubiquitin protein ligase binding
          alpha-tubulin binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 61736 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3337259 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.1 ST Array 7911309 - -
          Affymetrix Human Gene 1.1 ST Array 8159790 - -
          Affymetrix Human Gene 1.1 ST Array 8031981 - -
          Affymetrix Human Gene 1.1 ST Array 7986541 - -
          Affymetrix Human Gene 1.1 ST Array 8044649 - -
          Affymetrix Human Transcriptome Array 2.0 PSR19013355.hg.1 NR_033266.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19013355.hg.1 TC19000974.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC19000974.hg.1 NR_033266.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC19000974.hg.1 ENST00000391654.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC19000974.hg.1 NR_033266.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC19000974.hg.1 ENST00000391654.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC19000974.hg.1 ENST00000391654.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC19000974.hg.1 NR_033266.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19013357.hg.1 ENST00000391654.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19013357.hg.1 NR_033266.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19013357.hg.1 TC19000974.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19013358.hg.1 ENST00000391654.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19013358.hg.1 NR_033266.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19013358.hg.1 TC19000974.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1900171769.hg.1 ENST00000632506.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171769.hg.1 NR_033266.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900171769.hg.1 TC1900009065.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900171769.hg.1 ENST00000632506.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171769.hg.1 NR_033266.1 Refseq
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000631796.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000631796.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000631796.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000631796.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000631796.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000631796.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000631796.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000631796.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000631994.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000631994.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000631994.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000632089.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000632089.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000632089.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000632089.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000632089.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000632292.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000632292.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000632292.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000632292.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000632496.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000632496.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000632496.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000632496.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000632496.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000632506.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000632506.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000632506.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000632506.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000633703.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000633703.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000633703.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000633703.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000633719.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000633719.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000633719.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000633719.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000633719.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000633719.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000633719.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000633719.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000633719.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000633719.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000633719.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000633719.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000633719.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000633742.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000633742.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000633742.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000633742.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000634023.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 ENST00000634023.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 NR_033266.1 Refseq
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 NR_033266.1 Refseq
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 NR_033266.1 Refseq
          Affymetrix Human Clariom D Assay TC1900009065.hg.1 NR_033266.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900171784.hg.1 ENST00000631796.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171784.hg.1 ENST00000631994.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171784.hg.1 ENST00000632089.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171784.hg.1 ENST00000632506.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171784.hg.1 ENST00000633703.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171784.hg.1 ENST00000633719.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171784.hg.1 ENST00000633742.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171784.hg.1 NR_033266.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900171784.hg.1 TC1900009065.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900171784.hg.1 ENST00000631796.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171784.hg.1 ENST00000631994.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171784.hg.1 ENST00000632089.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171784.hg.1 ENST00000632506.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171784.hg.1 ENST00000633703.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171784.hg.1 ENST00000633719.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171784.hg.1 ENST00000633742.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171784.hg.1 NR_033266.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900171782.hg.1 ENST00000631796.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171782.hg.1 ENST00000631994.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171782.hg.1 ENST00000632089.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171782.hg.1 ENST00000632496.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171782.hg.1 ENST00000632506.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171782.hg.1 ENST00000633703.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171782.hg.1 ENST00000633719.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171782.hg.1 ENST00000633742.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171782.hg.1 NR_033266.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900171782.hg.1 TC1900009065.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900171782.hg.1 ENST00000631796.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171782.hg.1 ENST00000631994.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171782.hg.1 ENST00000632089.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171782.hg.1 ENST00000632496.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171782.hg.1 ENST00000632506.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171782.hg.1 ENST00000633703.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171782.hg.1 ENST00000633719.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171782.hg.1 ENST00000633742.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171782.hg.1 NR_033266.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900171780.hg.1 ENST00000631796.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171780.hg.1 ENST00000632089.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171780.hg.1 ENST00000632496.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171780.hg.1 ENST00000632506.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171780.hg.1 ENST00000633703.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171780.hg.1 ENST00000633719.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171780.hg.1 ENST00000633742.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171780.hg.1 ENST00000634023.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171780.hg.1 NR_033266.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900171780.hg.1 TC1900009065.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900171780.hg.1 ENST00000631796.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171780.hg.1 ENST00000632089.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171780.hg.1 ENST00000632496.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171780.hg.1 ENST00000632506.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171780.hg.1 ENST00000633703.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171780.hg.1 ENST00000633719.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171780.hg.1 ENST00000633742.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171780.hg.1 ENST00000634023.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900171780.hg.1 NR_033266.1 Refseq
          Human Genome U133 Plus 2.0 Array 225035_x_at - -
          Human Genome U133 Plus 2.0 Array 225995_PM_x_at - -
          Human Genome U133 Plus 2.0 Array 225995_x_at - -
          Human Genome U133 Plus 2.0 Array 225035_PM_x_at - -
          Human Genome U95 Set 54175_at - -
          Human Genome U95 Set 54175_PM_at - -
          Human Genome U95 Set 65766_PM_at - -
          Human Genome U95 Set 65766_at - -
          Human Genome U133 Set 225995_x_at - -
          Human Genome U133 Set 225995_PM_x_at - -
          Human Genome U133 Set 225035_x_at - -
          Human Genome U133 Set 225035_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A010783 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A010783 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1739199 BC063893.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1739199 BC094860.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1739199 BC063470.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1739199 BC094698.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1739199 BC053987.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1655952 CR621334.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1655952 CR621474.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1655952 BC063893.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1655952 BC094860.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1655952 BM970740.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3245020 CR621334.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3245020 CR621474.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3245020 BC063893.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3245020 BC094860.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3245020 BM970740.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1655952 CR621334.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1655952 CR621474.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1655952 BC063893.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1655952 BC094860.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1655952 BM970740.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b776c6558-ktt9x:80/100.66.134.75:80.
          git-commit: 08b7be5c3d3493be030128aff2ceec7cf49358c5
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc