Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05363036_g1
          See other LOC107985803 GE Assays ›
          Gene Symbol:
          LOC107985803 LINC00863 LOC642361
          Gene Name:
          uncharacterized LOC107985803
          long intergenic non-protein coding RNA 863
          uncharacterized LOC642361
          Gene Aliases:
          -
          -
          -
          Chromosome Location:
          Chr.2: 127858310 - 127887187 on Build GRCh38
          Chromosome Location:
          Chr.10: 87342411 - 87357302 on Build GRCh38
          Chromosome Location:
          Chr.10: 79825902 - 79827602 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          Assay ID Hs05363036_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC107985803
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_017005446.1 XP_016860935.1 5 2091 121
          Gene Symbol: LINC00863
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_029408.1 1 990 121
          GenBank mRNA CA444075.1 - 1 121
          CD369599.1 - 1 121
          Gene Symbol: LOC642361
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_029407.1 1 1451 119
          GenBank mRNA AF064843.1 - 1 1468 119
          BE044639.1 - 1 119
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AK025716.1 - 1 976 Chr. 2 - 127885997 - 127888809 [+] on Build GRCh38 121

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16369272 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27762436 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16369221 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27762415 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27762436 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16369221 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27762415 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16369272 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10006749.hg.1 NR_029408.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10006749.hg.1 TC10000616.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC10000616.hg.1 NR_029408.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC10000616.hg.1 NR_029408.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC10000616.hg.1 ENST00000439559.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC10000616.hg.1 NR_029408.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC10000616.hg.1 ENST00000439559.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC10000616.hg.1 ENST00000439559.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC10000616.hg.1 ENST00000439559.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC10000616.hg.1 ENST00000439559.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006748.hg.1 ENST00000439559.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10006748.hg.1 NR_029408.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10006748.hg.1 TC10000616.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR10006746.hg.1 NR_029408.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10006746.hg.1 TC10000616.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1000185839.hg.1 NR_029408.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000185839.hg.1 NR_029408.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000185839.hg.1 TC1000012473.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000185832.hg.1 NR_029408.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000185832.hg.1 NR_029408.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000185832.hg.1 TC1000012473.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000185834.hg.1 NR_029408.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000185834.hg.1 NR_029408.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000185834.hg.1 TC1000012473.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000185837.hg.1 NR_029408.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000185837.hg.1 NR_029408.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000185837.hg.1 TC1000012473.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000185836.hg.1 NR_029408.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000185836.hg.1 NR_029408.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000185836.hg.1 TC1000012473.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000185835.hg.1 NR_029408.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000185835.hg.1 NR_029408.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000185835.hg.1 TC1000012473.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC1000012473.hg.1 NR_029408.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012473.hg.1 NR_029408.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012473.hg.1 NR_029408.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012473.hg.1 NR_029408.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012473.hg.1 NR_029408.1 Refseq
          Affymetrix Human Clariom D Assay TC1000012473.hg.1 NR_029408.1 Refseq
          Human Genome U133 Plus 2.0 Array 228839_PM_s_at - -
          Human Genome U133 Plus 2.0 Array 228839_s_at - -
          Human Genome U95 Set 82148_PM_at - -
          Human Genome U95 Set 82148_at - -
          Human Genome U95 Set 70488_r_at - -
          Human Genome U95 Set 70488_PM_r_at - -
          Human Genome U133 Set 228842_PM_at - -
          Human Genome U133 Set 228839_s_at - -
          Human Genome U133 Set 236691_at - -
          Human Genome U133 Set 228839_PM_s_at - -
          Human Genome U133 Set 236691_PM_at - -
          Human Genome U133 Set 228842_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A044239 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A044239 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3218306 - -
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1743639 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1743639 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0