Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05612553_s1
          See other PCM1 GE Assays ›
          Gene Symbol:
          PCM1
          Gene Name:
          pericentriolar material 1
          Gene Aliases:
          PTC4, RET/PCM-1
          Chromosome Location:
          Chr.8: 17922763 - 18029948 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05612553_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PCM1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001315507.1 NP_001302436.1 39 7368 82
          NM_001315508.1 NP_001302437.1 36 6900 82
          NM_006197.3 NP_006188.3 39 7392 82
          XM_006716336.3 XP_006716399.1 42 7843 82
          XM_006716340.3 XP_006716403.1 42 7831 82
          XM_006716349.3 XP_006716412.1 38 7303 82
          XM_011544528.2 XP_011542830.1 42 7864 82
          XM_011544529.2 XP_011542831.1 44 8170 82
          XM_011544530.2 XP_011542832.1 43 7986 82
          XM_011544532.2 XP_011542834.1 42 7861 82
          XM_011544533.2 XP_011542835.1 42 7852 82
          XM_011544534.2 XP_011542836.1 42 7840 82
          XM_011544535.2 XP_011542837.1 42 7828 82
          XM_011544540.2 XP_011542842.1 42 7792 82
          XM_017013472.1 XP_016868961.1 43 7980 82
          XM_017013473.1 XP_016868962.1 43 7976 82
          XM_017013474.1 XP_016868963.1 40 7594 82
          XM_017013475.1 XP_016868964.1 42 7718 82
          XM_017013476.1 XP_016868965.1 39 7302 82
          XM_017013477.1 XP_016868966.1 42 7883 82
          XM_017013478.1 XP_016868967.1 42 7861 82
          XM_017013479.1 XP_016868968.1 42 7862 82
          XM_017013480.1 XP_016868969.1 40 7534 82
          XM_017013481.1 XP_016868970.1 42 7837 82
          XM_017013482.1 XP_016868971.1 39 7498 82
          XM_017013483.1 XP_016868972.1 41 7767 82
          XM_017013484.1 XP_016868973.1 38 7193 82
          XM_017013485.1 XP_016868974.1 41 7747 82
          XM_017013486.1 XP_016868975.1 39 7429 82
          XM_017013487.1 XP_016868976.1 39 7420 82
          XM_017013488.1 XP_016868977.1 39 7417 82
          XM_017013489.1 XP_016868978.1 41 7720 82
          XM_017013490.1 XP_016868979.1 41 7696 82
          XM_017013491.1 XP_016868980.1 38 7315 82
          XM_017013492.1 XP_016868981.1 38 7312 82
          XM_017013493.1 XP_016868982.1 40 7615 82
          XM_017013494.1 XP_016868983.1 38 7300 82
          XM_017013495.1 XP_016868984.1 40 7440 82
          XM_017013496.1 XP_016868985.1 37 7023 82
          XM_017013497.1 XP_016868986.1 38 7288 82
          XM_017013498.1 XP_016868987.1 38 7279 82
          XM_017013499.1 XP_016868988.1 40 7419 82
          XM_017013500.1 XP_016868989.1 38 7276 82
          XM_017013501.1 XP_016868990.1 40 7578 82
          XM_017013502.1 XP_016868991.1 37 6999 82

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PCM1

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          neuron migration
          interkinetic nuclear migration
          cytoplasmic microtubule organization
          microtubule anchoring
          microtubule anchoring at centrosome
          nonmotile primary cilium assembly
          intraciliary transport involved in cilium morphogenesis
          cilium assembly
          negative regulation of neurogenesis
          centrosome organization
          protein localization to centrosome
          positive regulation of intracellular protein transport
          neuronal stem cell population maintenance
          protein binding
          identical protein binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0