Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs06604664_s1
          See other LOC100129884 GE Assays ›
          Gene Symbol:
          LOC100129884
          Gene Name:
          uncharacterized LOC100129884
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs06604664_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100129884
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK097079.1 - 1 2381 128

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3395502 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P412613 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 12358 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 4525 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3395502 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P412613 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P412613 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array XM_001722013_at - -
          Affymetrix Human Gene 1.0 ST Array XM_001723940_at - -
          Affymetrix Human Gene 1.0 ST Array XM_001718836_at - -
          Affymetrix Human Gene 1.0 ST Array 7971966 - -
          Affymetrix Human Gene 1.1 ST Array 7971965 - -
          Affymetrix HT HG-U133+ PM Array Plate 234757_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 236560_PM_at - -
          Human Genome U133 Plus 2.0 Array 236560_PM_at - -
          Human Genome U133 Plus 2.0 Array 234757_at - -
          Human Genome U133 Plus 2.0 Array 234757_PM_at - -
          Human Genome U133 Plus 2.0 Array 236560_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234757_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234757_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234757_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234757_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236560_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236560_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234757_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234757_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236560_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236560_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236560_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236560_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234757_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234757_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236560_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234757_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236560_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234757_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236560_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236560_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 236560_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234757_at5 - -
          Human Genome U133 Set 234757_PM_at - -
          Human Genome U133 Set 236560_PM_at - -
          Human Genome U133 Set 234757_at - -
          Human Genome U133 Set 236560_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0