Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs06610609_s1
See other KMT2C GE Assays ›
Gene Symbol:
KMT2C
Gene Name:
lysine methyltransferase 2C
Gene Aliases:
HALR, MLL3
Chromosome Location:
Chr.7: 152134925 - 152436642 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs06610609_s1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: KMT2C
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_170606.2
NP_733751.2
52
13515
83
XM_005250025.4
XP_005250082.1
55
13534
83
XM_005250026.3
XP_005250083.1
55
13568
83
XM_005250027.4
XP_005250084.1
54
13739
83
XM_005250028.4
XP_005250085.1
54
13742
83
XM_005250031.4
XP_005250088.1
53
13577
83
XM_006716077.3
XP_006716140.1
54
13739
83
XM_006716078.3
XP_006716141.1
54
13670
83
XM_006716079.3
XP_006716142.1
53
13574
83
XM_011516450.2
XP_011514752.1
53
13694
83
XM_011516451.2
XP_011514753.1
53
13622
83
XM_011516452.2
XP_011514754.1
52
13589
83
XM_011516453.2
XP_011514755.1
53
13505
83
XM_011516454.2
XP_011514756.1
49
12737
83
XM_011516456.2
XP_011514758.1
53
13694
83
XM_017012480.1
XP_016867969.1
55
13571
83
XM_017012481.1
XP_016867970.1
55
13531
83
XM_017012482.1
XP_016867971.1
54
13739
83
XM_017012483.1
XP_016867972.1
54
13739
83
XM_017012484.1
XP_016867973.1
54
13709
83
XM_017012485.1
XP_016867974.1
53
13691
83
XM_017012486.1
XP_016867975.1
54
13667
83
XM_017012487.1
XP_016867976.1
54
13710
83
XM_017012488.1
XP_016867977.1
53
13559
83
XM_017012489.1
XP_016867978.1
35
10249
83
XM_017012490.1
XP_016867979.1
32
9821
83
GenBank mRNA
AF264750.2
-
39
11013
83
AL832387.1
-
16
5978
83
AL833924.1
-
9
1662
83
AY024361.1
-
52
13476
83
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length between 71 and 85
Detect Genomic DNA
Homo sapiens
Both primers and probe map within a single exon
Matching Ambion siRNAs:
Click here to find siRNAs for Gene KMT2C
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
histone H3-K4 methylation
DNA binding
protein binding
zinc ion binding
transferase activity, transferring acyl groups
histone-lysine N-methyltransferase activity
histone methyltransferase activity (H3-K4 specific)
poly(A) RNA binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0