Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs06628759_s1
          See other TRIT1 GE Assays ›
          Gene Symbol:
          TRIT1
          Gene Name:
          tRNA isopentenyltransferase 1
          Gene Aliases:
          GRO1, IPPT, IPT, IPTase, MOD5, hGRO1
          Chromosome Location:
          Chr.1: 39841031 - 39883511 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs06628759_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TRIT1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001312691.1 NP_001299620.1 1 124 80
          NM_001312692.1 NP_001299621.1 1 124 80
          NM_017646.5 NP_060116.2 1 124 80
          NR_132401.1 1 124 80
          NR_132402.1 1 124 80
          NR_132403.1 1 124 80
          NR_132404.1 1 124 80
          NR_132405.1 1 124 80
          NR_132406.1 1 124 80
          NR_132407.1 1 124 80
          NR_132408.1 1 124 80
          NR_132409.1 1 124 80
          NR_132410.1 1 124 80
          NR_132412.1 1 124 80
          NR_132413.1 1 124 80
          NR_132414.1 1 124 80
          NR_132415.1 1 124 80
          XM_006710706.1 XP_006710769.1 1 117 80
          GenBank mRNA AF074918.1 - 1 118 80
          AK223478.1 - 1 117 80
          AK296453.1 - 1 109 80
          AK304759.1 - 1 117 80
          AK307910.1 - 1 109 80
          AY052768.1 - 1 86 80
          AY303390.1 - 1 118 80
          AY702933.1 - 1 47 80
          AY702934.1 - 1 47 80
          AY702935.1 - 1 47 80
          AY702936.1 - 1 47 80
          AY702937.1 - 1 47 80
          AY702938.1 - 1 47 80
          AY702939.1 - 1 47 80
          AY702940.1 - 1 47 80
          AY702941.1 - 1 47 80
          AY702942.1 - 1 47 80
          AY702943.1 - 1 47 80
          AY702944.1 - 1 47 80
          AY702945.1 - 1 47 80
          AY702946.1 - 1 47 80
          AY702947.1 - 1 47 80
          BC010741.1 - 1 86 80
          BC107569.1 - 1 123 4594069 80
          BC128155.1 - 1 118 40112695 80
          BC128156.1 - 1 118 40112698 80
          BE294565.1 - 1 125 81
          DA031689.1 - 1 119 80

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AY702933.1, AY702934.1, AY702935.1, AY702936.1, AY702937.1, AY702938.1, AY702939.1, AY702940.1, AY702941.1, AY702942.1, AY702943.1, AY702944.1, AY702945.1, AY702946.1, XM_006710706.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TRIT1

          Panther Classification:

          Molecular Function -

          RNA processing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA modification
          mitochondrial tRNA modification
          nucleic acid binding
          ATP binding
          zinc ion binding
          tRNA dimethylallyltransferase activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0