Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs06638908_s1
          See other ABI3BP GE Assays ›
          Gene Symbol:
          ABI3BP
          Gene Name:
          ABI family member 3 binding protein
          Gene Aliases:
          NESHBP, TARSH
          Chromosome Location:
          Chr.3: 100749335 - 100993508 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs06638908_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ABI3BP
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_015429.3 NP_056244.2 2 229 97
          XM_005247281.2 XP_005247338.1 2 232 97
          XM_005247282.2 XP_005247339.1 2 232 97
          XM_005247283.2 XP_005247340.1 2 232 97
          XM_005247284.2 XP_005247341.1 2 232 97
          XM_005247285.2 XP_005247342.1 2 232 97
          XM_005247287.2 XP_005247344.1 2 232 97
          XM_005247288.2 XP_005247345.1 2 232 97
          XM_005247289.2 XP_005247346.1 2 232 97
          XM_005247290.2 XP_005247347.1 2 232 97
          XM_005247291.2 XP_005247348.1 2 232 97
          XM_005247293.2 XP_005247350.1 2 232 97
          XM_005247294.2 XP_005247351.1 2 232 97
          XM_005247295.2 XP_005247352.1 2 232 97
          XM_005247296.2 XP_005247353.1 2 232 97
          XM_005247298.2 XP_005247355.1 2 232 97
          XM_005247299.3 XP_005247356.1 2 247 97
          XM_005247300.2 XP_005247357.1 2 232 97
          XM_005247301.2 XP_005247358.1 2 232 97
          XM_005247302.2 XP_005247359.1 2 232 97
          XM_005247303.2 XP_005247360.1 2 232 97
          XM_005247305.2 XP_005247362.1 2 232 97
          XM_005247306.2 XP_005247363.1 2 232 97
          XM_006713569.2 XP_006713632.1 2 232 97
          XM_006713570.2 XP_006713633.1 2 232 97
          XM_006713571.2 XP_006713634.1 2 232 97
          XM_006713572.2 XP_006713635.1 2 232 97
          XM_011512639.1 XP_011510941.1 2 232 97
          XM_011512640.1 XP_011510942.1 2 232 97
          XM_011512641.1 XP_011510943.1 2 232 97
          XM_011512643.1 XP_011510945.1 2 232 97
          XM_011512644.1 XP_011510946.1 2 232 97
          XM_011512645.1 XP_011510947.1 2 232 97
          XM_011512646.1 XP_011510948.1 2 232 97
          XM_011512647.1 XP_011510949.1 2 232 97
          XM_011512648.1 XP_011510950.1 2 232 97
          XM_011512649.1 XP_011510951.1 2 232 97
          XM_011512651.1 XP_011510953.1 2 232 97
          XM_011512652.1 XP_011510954.1 2 232 97
          XM_011512653.1 XP_011510955.1 2 232 97
          XM_011512654.1 XP_011510956.1 2 232 97
          XM_011512656.1 XP_011510958.1 2 232 97
          XM_011512657.1 XP_011510959.1 2 232 97
          XM_011512658.1 XP_011510960.1 2 232 97
          XM_011512659.1 XP_011510961.1 2 232 97
          XM_011512660.1 XP_011510962.1 2 232 97
          XM_017006105.1 XP_016861594.1 2 232 97
          XM_017006106.1 XP_016861595.1 2 232 97
          XM_017006107.1 XP_016861596.1 2 232 97
          XM_017006108.1 XP_016861597.1 2 232 97
          GenBank mRNA AK025204.1 - 2 189 97
          AL833204.1 - 2 192 97
          BC030221.1 - 2 209 5205716 97
          DA342832.1 - 2 229 97

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene ABI3BP

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cell-substrate adhesion
          extracellular matrix organization
          collagen binding
          heparin binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0