Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs06644121_s1
          See other GNAS GE Assays ›
          Gene Symbol:
          GNAS
          Gene Name:
          GNAS complex locus
          Gene Aliases:
          AHO, C20orf45, GNAS1, GPSA, GSA, GSP, NESP, POH, SCG6, SgVI
          Chromosome Location:
          Chr.20: 58839681 - 58911196 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs06644121_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GNAS
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000516.5 NP_000507.1 13 1869 107
          NM_001077488.3 NP_001070956.1 14 1872 107
          NM_001077489.3 NP_001070957.1 12 1824 107
          NM_001077490.2 NP_001070958.1 13 3659 107
          NM_001309840.1 NP_001296769.1 13 1535 107
          NM_001309861.1 NP_001296790.1 13 1580 107
          NM_016592.3 NP_057676.1 14 2478 107
          NM_080425.3 NP_536350.2 13 3659 107
          NM_080426.3 NP_536351.1 13 1827 107
          XM_017027812.1 XP_016883301.1 14 3929 107
          XM_017027813.1 XP_016883302.1 13 3884 107
          XM_017027814.1 XP_016883303.1 12 3881 107
          XM_017027815.1 XP_016883304.1 12 1352 107
          XM_017027816.1 XP_016883305.1 14 2157 107
          XM_017027817.1 XP_016883306.1 12 1439 107
          XM_017027818.1 XP_016883307.1 12 1362 107
          XM_017027819.1 XP_016883308.1 12 1403 107
          XM_017027820.1 XP_016883309.1 12 1343 107
          GenBank mRNA AF105253.1 - 14 2456 107
          AK054862.1 - 11 2733 107
          AK093534.1 - 12 1679 107
          AK122771.1 - 11 1852 107
          BC008855.2 - 15 1590 107
          BC022875.1 - 12 1291 107
          BC066923.1 - 12 1403 107
          BC108315.1 - 12 1665 107
          BQ029639.1 - 1 107
          CA335150.1 - 1 169 107
          X04408.1 - 13 1513 107
          X04409.1 - 13 1415 107
          X07036.1 - 13 1455 107
          X56009.1 - 13 1521 107
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA Z70769.1 - 1 Chr. 20 - 58910969 - 58911186 [-] on Build GRCh38 107

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene GNAS

          Panther Classification:

          Molecular Function -

          heterotrimeric G-protein

          Gene Ontology Categories:

          Function(s) Process(es)

          tissue homeostasis
          endochondral ossification
          energy reserve metabolic process
          DNA methylation
          adenylate cyclase-activating G-protein coupled receptor signaling pathway
          adenylate cyclase-activating dopamine receptor signaling pathway
          sensory perception of chemical stimulus
          embryonic hindlimb morphogenesis
          multicellular organism growth
          post-embryonic body morphogenesis
          response to drug
          positive regulation of osteoblast differentiation
          positive regulation of osteoclast differentiation
          developmental growth
          embryonic cranial skeleton morphogenesis
          cognition
          cartilage development
          bone development
          hair follicle placode formation
          platelet aggregation
          genetic imprinting
          regulation of parathyroid hormone secretion
          GTPase activity
          signal transducer activity
          insulin-like growth factor receptor binding
          protein binding
          GTP binding
          G-protein beta/gamma-subunit complex binding
          beta-2 adrenergic receptor binding
          D1 dopamine receptor binding
          mu-type opioid receptor binding
          ionotropic glutamate receptor binding
          metal ion binding
          corticotropin-releasing hormone receptor 1 binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0