Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs07288798_m1
          See other AGO2 GE Assays ›
          Gene Symbol:
          AGO2
          Gene Name:
          argonaute 2, RISC catalytic component
          Gene Aliases:
          EIF2C2, Q10
          Chromosome Location:
          Chr.8: 140520158 - 140642406 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs07288798_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AGO2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164623.1 NP_001158095.1 7 - 8 918 88
          NM_012154.3 NP_036286.2 7 - 8 918 88
          XM_011516965.2 XP_011515267.1 7 - 8 3994 88
          XM_011516966.2 XP_011515268.1 7 - 8 1735 88
          XM_011516968.2 XP_011515270.2 7 - 8 1069 88
          XM_017013317.1 XP_016868806.1 6 - 7 687 88
          GenBank mRNA AY077717.1 - 5 - 6 856 88
          BC018727.1 - 2 - 3 165 88
          BC125213.1 - 6 - 7 724 88

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene AGO2

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          translation
          translational initiation
          Wnt signaling pathway, calcium modulating pathway
          post-embryonic development
          RNA secondary structure unwinding
          miRNA metabolic process
          gene silencing by RNA
          pre-miRNA processing
          siRNA loading onto RISC involved in RNA interference
          post-transcriptional gene silencing by RNA
          production of miRNAs involved in gene silencing by miRNA
          miRNA mediated inhibition of translation
          mRNA cleavage involved in gene silencing by miRNA
          miRNA loading onto RISC involved in gene silencing by miRNA
          positive regulation of transcription by RNA polymerase II
          negative regulation of translational initiation
          phosphatidylinositol-mediated signaling
          positive regulation of nuclear-transcribed mRNA poly(A) tail shortening
          RNA phosphodiester bond hydrolysis, endonucleolytic
          mRNA cleavage involved in gene silencing by siRNA
          positive regulation of nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
          RNA 7-methylguanosine cap binding
          RNA polymerase II complex binding
          core promoter sequence-specific DNA binding
          RNA binding
          double-stranded RNA binding
          single-stranded RNA binding
          mRNA binding
          translation initiation factor activity
          endoribonuclease activity
          protein binding
          protein C-terminus binding
          siRNA binding
          miRNA binding
          metal ion binding
          endoribonuclease activity, cleaving siRNA-paired mRNA
          endoribonuclease activity, cleaving miRNA-paired mRNA

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0