Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs99999177_g1
          See other PCNA GE Assays ›
          Gene Symbol:
          PCNA
          Gene Name:
          proliferating cell nuclear antigen
          Gene Aliases:
          ATLD2
          Chromosome Location:
          Chr.20: 5114953 - 5126622 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs99999177_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PCNA
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002592.2 NP_002583.1 2 - 3 458 69
          NM_182649.1 NP_872590.1 1 - 2 422 69
          GenBank mRNA AK300558.1 - 2 - 3 377 69
          AK313286.1 - 1 - 2 379 69
          BC000491.2 - 1 - 2 364 2820036 69
          BC062439.1 - 1 - 2 379 4102284 69
          BG612192.1 - 1 - 2 424 69
          CR536501.1 - 1 - 2 219 69
          CR541799.1 - 1 - 2 219 69
          M15796.1 - 1 - 2 337 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2000008279.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC20000592.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PCNA

          Panther Classification:

          Molecular Function -

          DNA polymerase processivity factor

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          regulation of transcription involved in G1/S transition of mitotic cell cycle
          telomere maintenance via recombination
          telomere maintenance
          DNA strand elongation involved in DNA replication
          leading strand elongation
          transcription-coupled nucleotide-excision repair
          nucleotide-excision repair, DNA incision, 5'-to lesion
          nucleotide-excision repair, DNA gap filling
          mismatch repair
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          heart development
          cell proliferation
          protein sumoylation
          translesion synthesis
          epithelial cell differentiation
          replication fork processing
          positive regulation of deoxyribonuclease activity
          nucleotide-excision repair, DNA incision
          response to lipid
          cellular response to UV
          error-prone translesion synthesis
          DNA damage response, detection of DNA damage
          positive regulation of DNA repair
          positive regulation of DNA replication
          response to cadmium ion
          error-free translesion synthesis
          mitotic telomere maintenance via semi-conservative replication
          purine-specific mismatch base pair DNA N-glycosylase activity
          chromatin binding
          damaged DNA binding
          protein binding
          enzyme binding
          estrogen receptor binding
          DNA polymerase processivity factor activity
          receptor tyrosine kinase binding
          dinucleotide insertion or deletion binding
          MutLalpha complex binding
          histone acetyltransferase binding
          identical protein binding
          DNA polymerase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 ENST00000379143.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 ENST00000379143.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 ENST00000379143.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 ENST00000379143.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 ENST00000379143.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 ENST00000379143.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 ENST00000379143.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 ENST00000379143.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 ENST00000379143.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 ENST00000379143.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 ENST00000379143.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 ENST00000379143.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 ENST00000379160.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 ENST00000379160.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 ENST00000379160.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 ENST00000379160.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 ENST00000379160.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 ENST00000379160.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 ENST00000379160.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 ENST00000379160.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 ENST00000379160.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 ENST00000379160.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 NM_002592.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 NM_002592.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 NM_002592.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 NM_002592.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 NM_002592.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 NM_002592.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 NM_002592.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 NM_002592.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 NM_002592.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 NM_002592.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 NM_182649.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 NM_182649.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 NM_182649.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 NM_182649.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 NM_182649.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 NM_182649.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 NM_182649.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 NM_182649.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 NM_182649.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 NM_182649.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000008279.hg.1 NM_182649.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20008653.hg.1 ENST00000379143.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20008653.hg.1 NM_182649.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20008653.hg.1 TC20000592.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR20008655.hg.1 ENST00000379143.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR20008655.hg.1 NM_182649.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20008655.hg.1 TC20000592.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0X00009520.hg.1 ENST00000397222.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2000158376.hg.1 ENST00000379143.9 Ensembl
          Affymetrix Human Clariom D Assay PSR2000158376.hg.1 NM_182649.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000158376.hg.1 TC2000008279.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000158376.hg.1 ENST00000379143.9 Ensembl
          Affymetrix Human Clariom D Assay PSR2000158376.hg.1 NM_182649.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000158379.hg.1 ENST00000379143.9 Ensembl
          Affymetrix Human Clariom D Assay PSR2000158379.hg.1 NM_182649.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000158379.hg.1 TC2000008279.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000158379.hg.1 ENST00000379143.9 Ensembl
          Affymetrix Human Clariom D Assay PSR2000158379.hg.1 NM_182649.1 Refseq
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 ENST00000379143.9 Ensembl
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 ENST00000379143.9 Ensembl
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 ENST00000379143.9 Ensembl
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 ENST00000379143.9 Ensembl
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 ENST00000379143.9 Ensembl
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 ENST00000379143.9 Ensembl
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 ENST00000379143.9 Ensembl
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 ENST00000379143.9 Ensembl
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 ENST00000379143.9 Ensembl
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 ENST00000379143.9 Ensembl
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 ENST00000379143.9 Ensembl
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 ENST00000379143.9 Ensembl
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 ENST00000379160.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 ENST00000379160.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 ENST00000379160.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 ENST00000379160.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 ENST00000379160.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 ENST00000379160.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 ENST00000379160.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 ENST00000379160.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 ENST00000379160.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 ENST00000379160.3 Ensembl
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 NM_002592.2 Refseq
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 NM_002592.2 Refseq
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 NM_002592.2 Refseq
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 NM_002592.2 Refseq
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 NM_002592.2 Refseq
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 NM_002592.2 Refseq
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 NM_002592.2 Refseq
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 NM_002592.2 Refseq
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 NM_002592.2 Refseq
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 NM_002592.2 Refseq
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 NM_182649.1 Refseq
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 NM_182649.1 Refseq
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 NM_182649.1 Refseq
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 NM_182649.1 Refseq
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 NM_182649.1 Refseq
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 NM_182649.1 Refseq
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 NM_182649.1 Refseq
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 NM_182649.1 Refseq
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 NM_182649.1 Refseq
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 NM_182649.1 Refseq
          Affymetrix Human Clariom D Assay TC2000008279.hg.1 NM_182649.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00009520.hg.1 ENST00000397222.2 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5756d5bff6-86w5v:80/100.66.130.62:80.
          git-commit: 27702359083d4c7b0962b032f2611a59cac079b5
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0