Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00433487_m1
          See other GABRG2 GE Assays ›
          Gene Symbol:
          Gabrg2
          Gene Name:
          gamma-aminobutyric acid (GABA) A receptor, subunit gamma 2
          Gene Aliases:
          GABAA-R, Gabrg-2, gamma2
          Chromosome Location:
          Chr.11: 41910190 - 42000840 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00433487_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gabrg2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008073.3 NP_032099.1 4 - 5 932 87
          NM_177408.6 NP_803127.3 4 - 5 932 87
          GenBank mRNA AF233775.1 - 4 - 5 548 87
          AF233776.1 - 4 - 5 548 87
          AF233777.1 - 4 - 5 548 87
          AF233778.1 - 4 - 5 548 87
          AF233779.1 - 4 - 5 548 87
          AF233780.1 - 4 - 5 548 87
          AF233781.1 - 4 - 5 548 87
          AF233782.1 - 4 - 5 548 87
          AF233783.1 - 4 - 5 548 87
          AF233784.1 - 4 - 5 548 87
          AF233785.1 - 4 - 5 548 87
          AF233786.1 - 4 - 5 548 87
          AF233787.1 - 4 - 5 548 87
          AF233788.1 - 4 - 5 548 87
          AF233789.1 - 4 - 5 548 87
          AF233790.1 - 4 - 5 548 87
          AF233791.1 - 4 - 5 548 87
          AF233792.1 - 4 - 5 548 87
          AF233793.1 - 4 - 5 548 87
          AF233794.1 - 4 - 5 548 87
          AF233795.1 - 4 - 5 548 87
          AF233796.1 - 4 - 5 548 87
          AF233797.1 - 4 - 5 548 87
          AF233798.1 - 4 - 5 548 87
          AF233799.1 - 4 - 5 548 87
          AF233800.1 - 4 - 5 548 87
          AF233801.1 - 4 - 5 548 87
          AF233802.1 - 4 - 5 548 87
          AK034338.1 - 4 - 5 902 87
          AK047085.1 - 4 - 5 510 87
          AK049857.1 - 4 - 5 575 87
          AK136889.1 - 4 - 5 540 87
          AK141129.1 - 4 - 5 574 87
          BC031762.1 - 4 - 5 750 4504205 87
          M62374.1 - 4 - 5 895 87
          M86572.1 - 4 - 5 724 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002553.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gabrg2

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          chloride transport
          signal transduction
          gamma-aminobutyric acid signaling pathway
          chemical synaptic transmission
          post-embryonic development
          adult behavior
          synaptic transmission, GABAergic
          cellular response to histamine
          chloride transmembrane transport
          GABA-A receptor activity
          extracellular ligand-gated ion channel activity
          chloride channel activity
          protein binding
          GABA receptor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100023638.mm.1 TC1100002553.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100023638.mm.1 NM_008073.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100023638.mm.1 NM_177408.6 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix