Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00433791_g1
          See other GRIN1 GE Assays ›
          Gene Symbol:
          Grin1
          Gene Name:
          glutamate receptor, ionotropic, NMDA1 (zeta 1)
          Gene Aliases:
          GluN1, GluRdelta1, GluRzeta1, M100174, NMD-R1, NMDAR1, NR1, Nmdar, Rgsc174
          Chromosome Location:
          Chr.2: 25291177 - 25319198 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00433791_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Grin1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001177656.2 NP_001171127.1 10 - 11 1703 97
          NM_001177657.2 NP_001171128.1 9 - 10 1640 97
          NM_008169.3 NP_032195.1 9 - 10 1640 97
          GenBank mRNA AK158140.1 - 9 - 10 1580 97
          AK158175.1 - 10 - 11 1641 97
          BC039157.1 - 9 - 10 1556 4507986 97
          D10028.1 - 9 - 10 1432 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003142.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Grin1

          Gene Ontology Categories:

          Function(s) Process(es)

          conditioned taste aversion
          startle response
          suckling behavior
          response to amphetamine
          transport
          ion transport
          cation transport
          calcium ion transport
          cellular calcium ion homeostasis
          chemical synaptic transmission
          respiratory gaseous exchange by respiratory system
          learning or memory
          learning
          memory
          long-term memory
          associative learning
          adult locomotory behavior
          olfactory learning
          visual learning
          regulation of cell communication
          positive regulation of cell death
          propylene metabolic process
          sensory perception of pain
          pons maturation
          cerebral cortex development
          ion transmembrane transport
          regulation of ion transmembrane transport
          social behavior
          ionotropic glutamate receptor signaling pathway
          synaptic transmission, glutamatergic
          regulation of membrane potential
          positive regulation of apoptotic process
          response to morphine
          regulation of neuron apoptotic process
          negative regulation of neuron apoptotic process
          regulation of respiratory gaseous exchange
          response to ethanol
          positive regulation of transcription by RNA polymerase II
          regulation of synaptic plasticity
          regulation of neuronal synaptic plasticity
          regulation of long-term neuronal synaptic plasticity
          rhythmic process
          regulation of dendrite morphogenesis
          regulation of axonogenesis
          neuromuscular process
          protein tetramerization
          regulation of synapse assembly
          calcium ion homeostasis
          excitatory postsynaptic potential
          prepulse inhibition
          male mating behavior
          calcium ion transmembrane transport
          cation transmembrane transport
          positive regulation of excitatory postsynaptic potential
          ionotropic glutamate receptor activity
          NMDA glutamate receptor activity
          signaling receptor binding
          ion channel activity
          extracellularly glutamate-gated ion channel activity
          cation channel activity
          calcium channel activity
          calcium ion binding
          protein binding
          calmodulin binding
          glycine binding
          glutamate binding
          enzyme binding
          voltage-gated cation channel activity
          glutamate receptor binding
          signaling receptor activity
          neurotransmitter binding
          protein heterodimerization activity
          protein dimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200025900.mm.1 TC0200003142.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200025900.mm.1 NM_001177656.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200025900.mm.1 NM_001177657.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200025900.mm.1 NM_008169.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200025898.mm.1 TC0200003142.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200025898.mm.1 NM_001177656.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200025898.mm.1 NM_001177657.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200025898.mm.1 NM_008169.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200025901.mm.1 TC0200003142.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200025901.mm.1 NM_001177656.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200025901.mm.1 NM_001177657.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200025901.mm.1 NM_008169.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200025899.mm.1 TC0200003142.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200025899.mm.1 NM_001177656.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200025899.mm.1 NM_001177657.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200025899.mm.1 NM_008169.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0