Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00436418_m1
          See other NR1H4 GE Assays ›
          Gene Symbol:
          Nr1h4
          Gene Name:
          nuclear receptor subfamily 1, group H, member 4
          Gene Aliases:
          AI957360, Fxr, HRR1, RIP14, Rxrip14
          Chromosome Location:
          Chr.10: 89454234 - 89533645 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00436418_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nr1h4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163700.1 NP_001157172.1 1 - 2 148 76
          NM_009108.2 NP_033134.2 1 - 2 148 76
          GenBank mRNA AK002513.1 - 1 - 2 135 76
          AK050566.1 - 1 - 2 129 76
          AK132255.1 - 1 - 2 140 76
          BC015261.1 - 1 - 2 119 4241705 76
          U09416.1 - 1 - 2 117 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002657.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nr1h4

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          nitrogen catabolite activation of transcription from RNA polymerase II promoter
          regulation of carbohydrate metabolic process
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          transcription by RNA polymerase II
          Notch signaling pathway
          bile acid metabolic process
          intracellular receptor signaling pathway
          regulation of urea metabolic process
          intracellular bile acid receptor signaling pathway
          steroid hormone mediated signaling pathway
          positive regulation of transcription by RNA polymerase II
          negative regulation of bile acid biosynthetic process
          cellular response to organonitrogen compound
          cellular response to bile acid
          positive regulation of glutamate metabolic process
          positive regulation of ammonia assimilation cycle
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          DNA-binding transcription factor activity, RNA polymerase II-specific
          DNA binding
          double-stranded DNA binding
          DNA-binding transcription factor activity
          steroid hormone receptor activity
          nuclear receptor activity
          zinc ion binding
          bile acid binding
          signaling receptor activity
          bile acid receptor activity
          peptide binding
          sequence-specific DNA binding
          metal ion binding
          retinoid X receptor binding
          chenodeoxycholic acid binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1000020236.mm.1 TC1000002657.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000020236.mm.1 NM_001163700.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000020236.mm.1 NM_001163700.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000020236.mm.1 NM_009108.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000020236.mm.1 NM_009108.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000020235.mm.1 TC1000002657.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000020235.mm.1 NM_001163504.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000020235.mm.1 NM_001163504.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000020235.mm.1 NM_001163700.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000020235.mm.1 NM_001163700.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000020235.mm.1 NM_009108.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000020235.mm.1 NM_009108.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix