Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00440365_g1
          See other MYH6 GE Assays ›
          Gene Symbol:
          Myh6
          Gene Name:
          myosin, heavy polypeptide 6, cardiac muscle, alpha
          Gene Aliases:
          A830009F23Rik, AA517445, Myhc-a, Myhca, alpha-MHC
          Chromosome Location:
          Chr.14: 54941921 - 54968014 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00440365_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Myh6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164171.1 NP_001157643.1 26 - 27 3935 104
          NM_010856.4 NP_034986.1 26 - 27 3871 104
          GenBank mRNA BC110700.1 - 26 - 27 3917 6786621 104
          BC138027.1 - 25 - 26 3810 8861046 104
          M76598.1 - 24 - 25 3733 104
          M76599.1 - 24 - 25 3733 104
          M76600.1 - 24 - 25 3733 104
          M76601.1 - 26 - 27 3835 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400002073.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Myh6

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          regulation of the force of heart contraction
          regulation of heart rate
          muscle contraction
          striated muscle contraction
          adult heart development
          visceral muscle development
          regulation of heart contraction
          metabolic process
          regulation of blood pressure
          actin filament-based movement
          muscle filament sliding
          myofibril assembly
          BMP signaling pathway
          regulation of ATPase activity
          sarcomere organization
          ATP metabolic process
          atrial cardiac muscle tissue morphogenesis
          ventricular cardiac muscle tissue morphogenesis
          cardiac muscle cell development
          cardiac muscle contraction
          canonical Wnt signaling pathway
          regulation of heart growth
          microfilament motor activity
          nucleotide binding
          cytoskeletal motor activity
          actin binding
          protein binding
          calmodulin binding
          ATP binding
          ATP hydrolysis activity
          protein kinase binding
          calcium-dependent ATPase activity
          identical protein binding
          protein homodimerization activity
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1400015683.mm.1 TC1400002073.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400015683.mm.1 NM_001164171.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015683.mm.1 NM_001164171.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015683.mm.1 NM_010856.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015683.mm.1 NM_010856.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015684.mm.1 TC1400002073.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400015684.mm.1 NM_001164171.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015684.mm.1 NM_001164171.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015684.mm.1 NM_010856.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015684.mm.1 NM_010856.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015685.mm.1 TC1400002073.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400015685.mm.1 NM_001164171.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015685.mm.1 NM_001164171.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015685.mm.1 NM_010856.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015685.mm.1 NM_010856.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015686.mm.1 TC1400002073.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400015686.mm.1 NM_001164171.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015686.mm.1 NM_001164171.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015686.mm.1 NM_010856.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015686.mm.1 NM_010856.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0