Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00440721_m1
          See other ABCB1B GE Assays ›
          Gene Symbol:
          Abcb1b
          Gene Name:
          ATP-binding cassette, sub-family B (MDR/TAP), member 1B
          Gene Aliases:
          Abcb1, Mdr1, Mdr1b, Pgy-1, Pgy1, mdr
          Chromosome Location:
          Chr.5: 8798147 - 8866315 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00440721_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Abcb1b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011075.2 NP_035205.1 5 - 6 481 93
          GenBank mRNA BC141363.1 - 5 - 6 438 9055975 93
          BC150811.1 - 5 - 6 438 9087722 93
          BQ829721.1 - 4 - 5 370 93
          M14757.1 - 5 - 6 441 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000052.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Abcb1b

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          xenobiotic transmembrane transport
          response to xenobiotic stimulus
          establishment of endothelial blood-brain barrier
          xenobiotic transport
          transmembrane transport
          nucleotide binding
          transporter activity
          ATP binding
          ABC-type xenobiotic transporter activity
          hydrolase activity
          ATP hydrolysis activity
          ATPase-coupled transmembrane transporter activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500000452.mm.1 TC0500000052.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500000452.mm.1 NM_011075.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000452.mm.1 NM_011075.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000454.mm.1 TC0500000052.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500000454.mm.1 NM_011075.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000454.mm.1 NM_011075.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000453.mm.1 TC0500000052.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500000453.mm.1 NM_011075.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000453.mm.1 NM_011075.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000451.mm.1 TC0500000052.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500000451.mm.1 NM_011075.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000451.mm.1 NM_011075.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000450.mm.1 TC0500000052.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500000450.mm.1 NM_011075.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500000450.mm.1 NM_011075.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0