Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00441265_m1
          See other CXCL15 GE Assays ›
          Gene Symbol:
          Cxcl15
          Gene Name:
          chemokine (C-X-C motif) ligand 15
          Gene Aliases:
          Il8, Scyb15, lungkine, weche
          Chromosome Location:
          Chr.5: 90794534 - 90803067 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00441265_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cxcl15
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011339.2 NP_035469.1 3 - 4 303 79
          GenBank mRNA AF082859.1 - 3 - 4 266 79
          AK142955.1 - 3 - 4 305 79
          BC061138.1 - 3 - 4 296 30298837 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000845.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cxcl15

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of leukocyte chemotaxis
          chemotaxis
          inflammatory response
          immune response
          G protein-coupled receptor signaling pathway
          hemopoiesis
          neutrophil chemotaxis
          response to lipopolysaccharide
          regulation of cell population proliferation
          chemokine-mediated signaling pathway
          cytokine activity
          chemokine activity
          CXCR chemokine receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P820196 NM_011339.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P820196 AK142955.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P820196 NM_011339.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P820196 AK142955.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 348359 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 967042 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 692381 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 140809 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 20049 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P128487 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P820196 NM_011339.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P820196 AK142955.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P820196 NM_011339.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P820196 AK142955.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P384187 NM_011339.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P384187 AF082859.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P384187 AK142955.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P384187 BC061138.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885261 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885259 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885267 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885253 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885260 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885243 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885230 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P128487 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05652281 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01158725 - -
          Whole Mouse Genome G4122A A_51_P384187 NM_011339.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P384187 AF082859.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P384187 AK142955.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P384187 BC061138.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P820196 NM_011339.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P820196 AK142955.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P820196 NM_011339.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P820196 AK142955.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114446 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119446 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P128487 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04813118 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04813114 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04813117 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04813115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04543037 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885249 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04898539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885269 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885233 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885237 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885243 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885235 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885287 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04201112 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885270 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885245 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885244 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04201426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04508537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885256 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04813118 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885231 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885240 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885230 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885283 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885229 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04205537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04898558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885281 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885242 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885260 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885241 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885228 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885266 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885257 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885276 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04706216 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10523150 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523148 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523147 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523149 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523146 - -
          Affymetrix HT MG-430 PM Array Plate 1421404_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1456428_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10523145 - -
          Mouse Genome 430 2.0 Array 1456428_at - -
          Mouse Genome 430 2.0 Array 1421404_PM_at - -
          Mouse Genome 430 2.0 Array 1421404_at - -
          Mouse Genome 430 2.0 Array 1456428_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20309_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20309_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500007435.mm.1 TC0500000845.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500007435.mm.1 NM_011339.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007435.mm.1 NM_011339.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007436.mm.1 TC0500000845.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500007436.mm.1 NM_011339.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007436.mm.1 NM_011339.2 Refseq
          Mouse Expression Set 430 1421404_PM_at - -
          Mouse Expression Set 430 1421404_at - -
          Mouse Expression Set 430 1456428_at - -
          Mouse Expression Set 430 1456428_PM_at - -
          Murine Genome U74v2 Set 165875_PM_at - -
          Murine Genome U74v2 Set 166136_PM_i_at - -
          Murine Genome U74v2 Set 166136_i_at - -
          Murine Genome U74v2 Set 165875_at - -
          Mouse Genome 430A 2.0 Array 1421404_PM_at - -
          Mouse Genome 430A 2.0 Array 1421404_at - -
          Affymetrix HT Mouse Genome 430A Array 1421404_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421404_at - -
          Affymetrix HT Mouse Genome 430B Array 1456428_at - -
          Affymetrix HT Mouse Genome 430B Array 1456428_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 570234 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2680451 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002680451 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2635462 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2635462 - -
          mouse-6 v1.1 (Illumina) scl27600.3.1_4-S - -
          Illumina Sentrix mouse-6.v1 bead array 2680451 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl27600.3.1_4-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2680451 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2635462 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2635462 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix