Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200034412.mm.1 |
TC0200004109.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200034412.mm.1 |
NM_001177730.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034412.mm.1 |
NM_001177730.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034412.mm.1 |
NM_013839.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034412.mm.1 |
NM_013839.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034420.mm.1 |
TC0200004109.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200034420.mm.1 |
NM_001177730.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034420.mm.1 |
NM_001177730.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034416.mm.1 |
TC0200004109.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200034416.mm.1 |
NM_001177730.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034416.mm.1 |
NM_001177730.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034416.mm.1 |
NM_013839.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200034416.mm.1 |
NM_013839.4 |
Refseq |