Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00444673_g1
          See other FXYD1 GE Assays ›
          Gene Symbol:
          Fxyd1
          Gene Name:
          FXYD domain-containing ion transport regulator 1
          Gene Aliases:
          0610012C17Rik, 1110006M24Rik, Plm, Pml
          Chromosome Location:
          Chr.7: 31051678 - 31055877 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00444673_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fxyd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019503.4 NP_062376.2 4 - 5 282 69
          NM_052991.4 NP_443717.1 4 - 5 265 69
          NM_052992.3 NP_443718.1 4 - 5 346 69
          NM_194321.2 NP_919302.1 4 - 5 329 69
          GenBank mRNA AA462069.1 - 4 - 5 214 69
          AA466595.1 - 4 - 5 282 69
          AF089734.1 - 4 - 5 208 69
          AK002585.1 - 4 - 5 269 69
          BC024671.1 - 4 - 5 219 4208962 69
          BY064152.1 - 4 - 5 281 69
          BY132051.1 - 4 - 5 251 69
          CA459152.1 - 4 - 5 287 69
          CK793316.1 - 4 - 5 264 69
          W16239.1 - 4 - 5 245 69
          W98524.1 - 4 - 5 260 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002788.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fxyd1

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          chloride transport
          positive regulation of organic acid transport
          regulation of cytosolic calcium ion concentration
          regulation of cardiac muscle cell contraction
          regulation of cardiac muscle cell membrane potential
          positive regulation of sodium ion export across plasma membrane
          positive regulation of inorganic anion transmembrane transport
          regulation of sodium ion transmembrane transporter activity
          ion channel activity
          chloride channel activity
          myosin binding
          sodium channel regulator activity
          ATPase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700024978.mm.1 TC0700002788.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024978.mm.1 NM_052991.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024978.mm.1 NM_052991.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024978.mm.1 NM_194321.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024978.mm.1 NM_194321.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024972.mm.1 TC0700002788.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024972.mm.1 NM_019503.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024972.mm.1 NM_019503.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024972.mm.1 NM_052991.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024972.mm.1 NM_052991.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024972.mm.1 NM_052992.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024972.mm.1 NM_052992.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024972.mm.1 NM_194321.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024972.mm.1 NM_194321.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024977.mm.1 TC0700002788.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024977.mm.1 NM_052992.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024977.mm.1 NM_052992.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024977.mm.1 NM_194321.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024977.mm.1 NM_194321.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024979.mm.1 TC0700002788.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024979.mm.1 NM_052991.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024979.mm.1 NM_052991.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024979.mm.1 NM_194321.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024979.mm.1 NM_194321.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024974.mm.1 TC0700002788.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024974.mm.1 NM_019503.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024974.mm.1 NM_019503.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024974.mm.1 NM_052991.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024974.mm.1 NM_052991.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024974.mm.1 NM_052992.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024974.mm.1 NM_052992.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024974.mm.1 NM_194321.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024974.mm.1 NM_194321.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-875ns:80/100.66.130.62:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0