Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00445261_m1
          See other UCN GE Assays ›
          Gene Symbol:
          Ucn
          Gene Name:
          urocortin
          Gene Aliases:
          Mpv17, Ucn1
          Chromosome Location:
          Chr.5: 31137989 - 31138895 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00445261_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ucn
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021290.2 NP_067265.1 1 - 2 103 60
          GenBank mRNA BC116948.1 - 1 - 2 37 40126267 60
          BC116950.1 - 1 - 2 37 40126269 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002174.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the BC116948.1, BC116950.1, NM_021290.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ucn

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          startle response
          response to oxidative stress
          neuropeptide signaling pathway
          sensory perception of sound
          learning or memory
          feeding behavior
          associative learning
          aerobic respiration
          positive regulation of gene expression
          negative regulation of gene expression
          response to auditory stimulus
          pancreatic juice secretion
          positive regulation of cell growth
          negative regulation of histone deacetylation
          neuron projection development
          negative regulation of appetite
          positive regulation of interleukin-6 production
          positive regulation of collagen biosynthetic process
          positive regulation of peptidyl-serine phosphorylation
          activation of protein kinase A activity
          social behavior
          gastric emptying
          response to immobilization stress
          response to starvation
          drinking behavior
          negative regulation of apoptotic process
          negative regulation of catalytic activity
          positive regulation of vascular permeability
          positive regulation of translation
          positive regulation of DNA replication
          negative regulation of blood pressure
          negative regulation of cell size
          positive regulation of transcription by RNA polymerase II
          negative regulation of hormone secretion
          positive regulation of corticotropin secretion
          regulation of synaptic transmission, glutamatergic
          prepulse inhibition
          positive regulation of cardiac muscle contraction
          negative regulation of gastric acid secretion
          negative regulation of necrotic cell death
          negative regulation of cell death
          positive regulation of calcium ion import
          negative regulation of neuron death
          negative regulation of feeding behavior
          positive regulation of behavioral fear response
          G protein-coupled receptor binding
          hormone activity
          histone deacetylase inhibitor activity
          corticotropin-releasing hormone receptor 1 binding
          corticotropin-releasing hormone receptor 2 binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23576666 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23576670 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23576667 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23576668 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23576669 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23576671 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2150208 NM_021290.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 688505 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153634 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2150208 NM_021290.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576685 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576668 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153634 - -
          Whole Mouse Genome G4122A A_51_P255952 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139809 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153634 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04977973 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04977959 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04908545 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04977981 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04977966 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04977963 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23576669 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23576671 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23576666 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23576668 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23576670 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23576667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576670 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576663 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04977972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576670 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576672 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576683 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23576680 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10529079 - -
          Affymetrix Mouse Gene 1.0 ST Array 10529080 - -
          Affymetrix Mouse Gene 1.0 ST Array 10529078 - -
          Affymetrix Mouse Gene 1.0 ST Array 10529081 - -
          Affymetrix HT MG-430 PM Array Plate 1450610_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10529077 - -
          Mouse Genome 430 2.0 Array 1450610_at - -
          Mouse Genome 430 2.0 Array 1450610_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22226_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22226_at - -
          Affymetrix Mouse Clariom S Array PSR0500019783.mm.1 TC0500002174.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500019783.mm.1 NM_021290.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500019783.mm.1 NM_021290.2 Refseq
          Mouse Expression Set 430 1450610_PM_at - -
          Mouse Expression Set 430 1450610_at - -
          Mouse Genome 430A 2.0 Array 1450610_PM_at - -
          Mouse Genome 430A 2.0 Array 1450610_at - -
          Affymetrix HT Mouse Genome 430A Array 1450610_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450610_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 324847 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6620338 - -
          Illumina mouse-6 v1.0 expression beadchip 6620338 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006620338 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1255017 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1255017 - -
          mouse-6 v1.1 (Illumina) scl26739.2.1_199-S - -
          Illumina Sentrix mouse-6.v1 bead array 6620338 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl26739.2.1_199-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6620338 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2834808 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2834808 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0